In Situ Cell Death Detection Kit, AP Protocol
Product No. 11684809910
Fixation of tissue sections
Roche recommends cross-linking fixatives, such as paraformaldehyde for the TUNEL assay (i.e., when using one of our In Situ Cell Death Detection Kits) to avoid that fragmented DNA of apoptotic cells is washed out of the cells during the assay procedure (which could result in false negatives). In the literature there are also TUNEL references using acetone- or methanol-based fixatives which do no cross-link cellular components. When using such fixatives it is possible that apoptosis is underestimated due to a loss of fragmented DNA. Fixation of cultured cells with the formalin-free Accustain® (Sigma) is as good as freshly prepared paraformaldehyde, but faster (fixation time is only 15 min compared to 60 min with paraformaldehyde). Alternatively, fixation with 2% glutaraldehyde can also be tested.
Preparation of 4% Paraformaldehyde (PFA)
In a screwcap bottle, mix 2 g PFA, 40 ml PBS and 100 μl 5N NaOH. Incubate at +56 °C in a water bath; shake periodically until PFA is completely dissolved. Set pH to 7.4 using 5N HCl (approx. 100 μl) and fill up to 50 ml final volume with PBS. Prepare this solution fresh, use it just after preparation, and discard the rest, according to local regulations for hazardous materials.
Using plastic embedded tissue sections
Plastic embedded tissue sections cannot be used with this kit. Polymerization of plastics used for tissue embedding (e.g., methyl methacrylate, glycol methacrylate, or epoxys like araldite and epon) is often induced by UV light. UV light also induces DNA strand breaks, and terminal transferase will label these strand breaks, resulting in false positive staining. For this reason, Roche does not recommend any embedding procedures which could induce DNA strand breaks for samples which are to be analyzed by the TUNEL method (i.e., when using one of our In Situ Cell Death Detection Kits).
Related Links