Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Klenow Enzyme Protocol
Product No. KLEN-RO
Protocol
Filling-in reaction
Protocols of standard assays for partial and complete filling in can be found in LabFaqs.
Alternative protocol:
  1. In a 20-μl reaction, digest 0.1 to 4 μg DNA with a restriction endonuclease.
  2. Add 1 μl of 0.5 mM of each dNTP.
    Note: It is usually not necessary to inactivate the restriction endonuclease, to change buffers, or to repurify the DNA prior to adding Klenow enzyme. If you nevertheless experience problems, try to adjust the buffer conditions to the SURE/Cut incubation Buffer M (f.c. 10 mM Tris, 10 mM MgCl ¬2, 50 mM NaCl, 1 mM DTT, pH 7.5).
  3. Add 1 to 5 U of Klenow enzyme and incubate at 30 °C for 15 min.
  4. Stop the reaction by heating to +75 °C for 10 min or by adding 1 μl of 0.5 M EDTA.
    For restriction fragments produced by cleavage with different endonucleases, it is possible to repair one end selectively. This is done by cleaving with enzyme 1, repairing the ends, inactivating the Klenow fragment by heat (+75 °C for 10 min), and cleaving with enzyme 2.
    Note: Repair of 5′ extensions is carried out by Klenow 's polymerase activity, whereas repair of 3′ extensions is carried out by 3′->5′exonuclease activity. Due to the relative inactivity of exonuclease, this method is not desirable in cases where extensive repair of overhanging 3′ ends is required. In such situations, T4 DNA polymerase is recommended.
Materials
SURE/CUT is a trademark of Roche
Related Links