Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Nick Translation Mix Protocol & Troubleshooting
Product No. 11745808910
Protocol
Protocol for chromosome spreads and DNA probes labeled via Nick translation
Reference: Chapter 5 of the Non-radioactive In situ Hybridization Manual (4th ed., p. 76ff).
Detection of human chromosomes in metaphase spreads using DIG or Fluorescein labeled Nick translated Human satellite probes: Reference can be found under the following link.
Troubleshooting
EFFICIENCY / No Signals with FISH
When your FISH shows no signals, first check probe length; if probe length is less than 200-500 bp, check if enough template (minimum 1 μg) was used for labeling. Determine if the protocol used corresponds to the guidelines in the package insert.
Create aliquots (2-3 portions) of the Nick translation Mix solution, because repeated freezing and thawing can degrade their activity.
Roche translation mixes are sensitive to the DNA concentration. Changing the recommended template amount can produce labeled fragments with an increasing size distribution. For this reason, it is important to accurately measure the template concentration, following the guidelines in the package insert of the DIG-Nick Translation Mix. The labeling volume should be 20 μl for each μg of template.