[Skip to Content](https://www.sigmaaldrich.com#main-content)
[](https://www.sigmaaldrich.com/US/en)
Products
Cart0
USEN
Products
ProductsApplicationsServicesResourcesSupport
[Login / Register](https://www.sigmaaldrich.com/oidc-sign-in)
[Order Lookup](https://www.sigmaaldrich.com/US/en/order-lookup)
[Quick Order](https://www.sigmaaldrich.com/US/en/quick-order)
Cart0
[Home](https://www.sigmaaldrich.com/US/en)[Plant Tissue Culture](https://www.sigmaaldrich.com/US/en/applications/cell-culture-and-cell-culture-analysis/cell-culture-by-cell-type/plant-tissue-culture)Silver Thiosulfate – Plant Tissue Culture Protocol
# Silver Thiosulfate – Plant Tissue Culture Protocol
Prepare a 0.1 M [Sodium Thiosulfate](https://www.sigmaaldrich.com/US/en/product/sigma/S7026) (STS) stock solution by dissolving 1.58 g of [Sodium Thiosulfate](https://www.sigmaaldrich.com/US/en/product/sigma/S7026) (Product No. [S7026](https://www.sigmaaldrich.com/US/en/product/sigma/S7026)) into 100 mL of water. Prepare a 0.1 M [Silver Nitrate](https://www.sigmaaldrich.com/US/en/product/sigma/S7276) stock solution by dissolving 1.7 g of [Silver Nitrate](https://www.sigmaaldrich.com/US/en/product/sigma/S7276) (Product No. [S7276](https://www.sigmaaldrich.com/US/en/product/sigma/S7276)) into 100 mL of water. Store the stock solutions in the dark until needed to prepare the STS.
In general, the STS is prepared with a molar ratio between silver and thiosulfate of 1:4, respectively. Nearly all of the silver present in the solution is in the form of \[Ag(S2O3)2]3-, the active complex for ethylene effect inhibition.
Prepare a 0.02 M STS by slowly pouring 20 mL of 0.1 M [silver nitrate](https://www.sigmaaldrich.com/US/en/product/sigma/S7276) stock solution into 80 mL of 0.1 M [sodium thiosulfate](https://www.sigmaaldrich.com/US/en/product/sigma/S7026) stock solution. The STS can be stored in the refrigerator for up to a month. However, preparation of the STS just prior to use is recommended.
## Materials
Sorry, an unexpected error has occurred
Response not successful: Received status code 500
__Related Articles__
- [Antibiotics in Plant Tissue Culture Protocol](https://www.sigmaaldrich.com/US/en/technical-documents/protocol/cell-culture-and-cell-culture-analysis/plant-tissue-culture/antibiotics)
- [Explant Sterilization – Plant Tissue Culture Protocol](https://www.sigmaaldrich.com/US/en/technical-documents/protocol/cell-culture-and-cell-culture-analysis/plant-tissue-culture/explant-sterilization)
- [Gelling Agents – Plant Tissue Culture Protocol](https://www.sigmaaldrich.com/US/en/technical-documents/protocol/cell-culture-and-cell-culture-analysis/plant-tissue-culture/gelling-agents)
- [Media Preparation – Plant Tissue Culture Protocol](https://www.sigmaaldrich.com/US/en/technical-documents/protocol/cell-culture-and-cell-culture-analysis/plant-tissue-culture/media-preparation)
- [Pathology Media – Plant Tissue Culture Protocol](https://www.sigmaaldrich.com/US/en/technical-documents/protocol/cell-culture-and-cell-culture-analysis/plant-tissue-culture/pathology-media)
- [Phycology Media in Plant Tissue Culture](https://www.sigmaaldrich.com/US/en/technical-documents/protocol/cell-culture-and-cell-culture-analysis/plant-tissue-culture/phycology-media)
- [Vitamin Mixtures – Plant Tissue Culture Protocol](https://www.sigmaaldrich.com/US/en/technical-documents/protocol/cell-culture-and-cell-culture-analysis/plant-tissue-culture/vitamin-mixtures)
- [Genome Editing in Plants with CRISPR/Cas9](https://www.sigmaaldrich.com/US/en/technical-documents/protocol/genomics/advanced-gene-editing/genome-editing-in-plants-with-crispr-cas9)
- [View More](https://www.sigmaaldrich.com/US/en/search/facet-search?focus=sitecontent&term=facet-search)
__Related Product Categories__
- [Salts](https://www.sigmaaldrich.com/US/en/products/chemistry-and-biochemicals/lab-chemicals/salts)
Top
__Sign In To Continue__
Sign In__Don't Have An Account?__Register
Support[Customer Support](https://www.sigmaaldrich.com/US/en/support/customer-support)[Contact Us](https://www.sigmaaldrich.com/US/en/collections/offices)[FAQ](https://maestro.my.site.com/knowledgeportal/s/)[Safety Data Sheets (SDS)](https://www.sigmaaldrich.com/US/en/documents-search?tab=sds)[Certificates (COA/COO)](https://www.sigmaaldrich.com/US/en/documents-search?tab=coa)[Quality & Regulatory](https://www.sigmaaldrich.com/US/en/life-science/quality-and-regulatory-management)[Calculators & Apps](https://www.sigmaaldrich.com/US/en/support/calculators-and-apps)[Webinars](https://www.sigmaaldrich.com/US/en/collections/webinars)
Orders[Order Lookup](https://www.sigmaaldrich.com/US/en/order-lookup)[Quick Order](https://www.sigmaaldrich.com/US/en/quick-order)[Custom Products](https://www.sigmaaldrich.com/US/en/services/custom-products)[eCommerce Solutions](https://www.sigmaaldrich.com/US/en/life-science/ecommerce/ecommerce-solutions)
Company[About Us](https://www.sigmaaldrich.com/US/en/life-science/about-us)[Responsibility](https://www.sigmaaldrich.com/US/en/life-science/ssbi)[Events](https://www.sigmaaldrich.com/US/en/collections/events)[Press Releases](https://www.sigmaaldrich.com/US/en/collections/press)[Programs](https://www.sigmaaldrich.com/US/en/life-science/partnership-programs)[Careers](https://careers.merckgroup.com/global/en)[Offices](https://www.sigmaaldrich.com/US/en/collections/offices)
Social Media
[](https://www.linkedin.com/company/merck-life-science)
[](https://x.com/Merck_lifesci)
[](https://www.facebook.com/Merck.lifescience)
[](https://www.instagram.com/mercklifescience/)
[](https://www.youtube.com/@MerckLifeScience/featured)
MilliporeSigma
__Research. Development. Production.__
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
[](https://www.sigmaaldrich.com/US/en)
[Sigma-Aldrich® Solutions](https://www.sigmaaldrich.com/US/en/life-science/sigma-aldrich)[BioReliance® Solutions](https://www.sigmaaldrich.com/US/en/life-science/bioreliance)[Millipore® Solutions](https://www.sigmaaldrich.com/US/en/life-science/millipore)[SAFC® Solutions](https://www.sigmaaldrich.com/US/en/life-science/safc)[Milli-Q® Solutions](https://www.sigmaaldrich.com/US/en/life-science/milliq)[Supelco® Solutions](https://www.sigmaaldrich.com/US/en/life-science/supelco)
© 2026 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved, including Text and Data Mining for AI training and similar technologies.
Reproduction of any materials from the site is strictly forbidden without permission.
[Site Use Terms](https://www.sigmaaldrich.com/US/en/life-science/legal/site-use-terms)|[Privacy Policy](https://www.sigmaaldrich.com/US/en/life-science/legal/privacy-statement)|[General Terms and Conditions of Sale](https://www.sigmaaldrich.com/US/en/life-science/legal/terms-and-conditions)|[Copyright Consent](https://www.sigmaaldrich.com/US/en/life-science/legal/copyright-consent) |[Site Map](https://www.sigmaaldrich.com/US/en/site-map)
An unknown error has occured.