[Skip to Content](https://www.sigmaaldrich.com#main-content)
[](https://www.sigmaaldrich.com/US/en)
Products
Cart0
USEN
Products
ProductsApplicationsServicesResourcesSupport
[Login / Register](https://www.sigmaaldrich.com/oidc-sign-in)
[Order Lookup](https://www.sigmaaldrich.com/US/en/order-lookup)
[Quick Order](https://www.sigmaaldrich.com/US/en/quick-order)
Cart0
[Home](https://www.sigmaaldrich.com/US/en)[Protein & Nucleic Acid Interactions](https://www.sigmaaldrich.com/US/en/applications/protein-biology/protein-and-nucleic-acid-interactions)Using Duolink™ in Multiwell Plates
# Using Duolink™ in Multiwell Plates
## Recommended modifications to the Duolink™ *In Situ* Fluorescence User Manual
When using the Duolink™ reagents to stain cells grown in multiwell plates, a few modifications to the Duolink™ protocol, optimized for staining samples deposited on glass slides, have to be made in order to suit the multiwell format. In particular, the washing procedure, the preparation for imaging, and the plate scanning differ from the standard protocol. We recommend you carefully read through the Duolink™ *In Situ* Fluorescence User Manual and adjust the protocol according to the following guidelines when staining cells in 96-well multiwell plates. Additional modifications to the protocol might also be necessary to meet your specific application or assay design.
## Recommended Equipment
- PerkinElmer® ViewPlate-96 Black, Optically Clear Bottom (PerkinElmer, Art no. 6005182, case of 50) or similar plates
- Plastic multiwell plate lid, adhesive plastic film or Parafilm®
- Orbital shakers for incubation and wash steps
- Cellomics ArrayScan or other high resolution laser scanning microscope
- Duolink™ *In situ* Microplate Nuclear Stain and Anti-Fade ([DUO82064](https://www.sigmaaldrich.com/US/en/product/SIGMA/DUO82064))
- Duolink™ *In situ* Microplate Heat Transfer Block ([DUO82065](https://www.sigmaaldrich.com/US/en/product/SIGMA/DUO82065))
## Pre-Treatment, Blocking, and Primary Antibodies
Grow cells in multiwell plates and treat (starve, stimulate etc.), wash, and fix the cells according to your standard immunostaining protocol. The conditions for your primary antibodies should be optimized with respect to sample fixation, antigen retrieval, blocking solution, antibody diluent, concentrations, and incubation temperature and time.
## Incubation Steps
40 μl of reagents per well is recommended. During the incubations, cover the plate with a plastic plate lid, adhesive plastic film, or Parafilm to prevent drying of the samples. Incubation in a pre-heated Microplate Heat Transfer Block will increase staining efficiency and reproducibility between different plates and reduce edge effects. It is also recommended to place the plate on an orbital shaker (maximum 240 rpm) during the incubation steps to improve the evenness of the staining.
## Wash Steps
At least 200 μl of 1x Wash Buffer A per well is recommended per wash step. Washing can be done either manually or automatic with a multiwell cell washer module. Try to remove as much remaining wash buffer before adding new reagents to avoid interference. At the same time, do not allow the samples to dry and make sure the washing is not too violent since the cells may detach. After each wash step, let the samples soak in wash buffer before aspirating, e.g. during a 2 x 5 min wash: wash with 200 μl and let soak for 5 min, aspirate, then wash in another 200 μl and let soak for 5 min, aspirate and add new reagents. During the Final Wash Step in Wash Buffer B, let the samples soak for 2 x 10 min in 1x Wash Buffer B, before proceeding to preparation for imaging.
## Preparation for imaging
For nuclear staining with DAPI and preserving PLA signals, use Microplate Nuclear Stain and Anti-Fade which contains Nuclear Staining Buffer (10x) and Anti- Fade Buffer (10x). Start this protocol after the Final Wash Step in 1x Wash Buffer B for 2 x 10 min:
1. Bring the Nuclear Staining Buffer to room temperature.
2. Add 9.0 mL of high purity water to the vial to obtain a 1x Nuclear Staining Buffer.
3. Let the samples soak in 1x Wash Buffer A for 2 min.
4. Remove the wash buffer and add 50 μl/well of 1x Nuclear Staining Buffer.
5. Incubate for 30 min at 37 °C. In the end of this incubation bring the Anti-Fade Buffer to room temperature.
6. Add 9.0 mL of high purity water to the vial to obtain a 1x Anti- Fade Buffer.
7. Let the samples soak in 1x Wash Buffer A for 2 min.
8. Remove the wash buffer and add 50 μl/well of 1x Anti-Fade Buffer.
9. Scan the plate.
To prevent evaporation during prolonged scanning, seal the plate with an adhesive plastic film. Scanned plates should be stored at 4 °C, protected from light. The signals will remain for a week if stored at 4 °C.
## Plate Scanning
Use scanning settings appropriate for your microscope and make sure the correct filters are selected for image acquisition. At least 20x magnification should be used. A fixed exposure time in the signal channel is required to be able to compare data between wells and scanning should preferably be done at a resolution of 0.25 μm/pixel but at least 0.45 μm/pixel.
## Recommendations for 384-Well Plates
Use PerkinElmer® ViewPlate-384 Black, Optically Clear Bottom (PerkinElmer, Art no. 6007460, case of 40) or similar plates and at least 15 μl of reagents per well. Be careful with washings since bubbles easily form, using TBS without TWEEN® 20 may help. Bubbles can also be removed by centrifuging the plate briefly. Uneven staining across the plate can be avoided by putting the plate on ice during the pipetting steps and then put the plate in a pre-heated Microplate Heat Transfer Block during the 37 °C incubations.
### References
1\.
Leuchowius K, Jarvius M, Wickström M, Rickardson L, Landegren U, Larsson R, Söderberg O, Fryknäs M, Jarvius J. 2010. High Content Screening for Inhibitors of Protein Interactions and Post-translational Modifications in Primary Cells by Proximity Ligation. Mol Cell Proteomics. 9(1):178-183. [https://doi.org/10.1074/mcp.m900331-mcp200](https://doi.org/10.1074/mcp.m900331-mcp200)
__Related Articles__
- [RNA Immunoprecipitation qPCR (RIP- qPCR) Protocol](https://www.sigmaaldrich.com/US/en/technical-documents/protocol/genomics/qpcr/rip-qpcr-protocol)
- [Northern & Southern Blot Protocols](https://www.sigmaaldrich.com/US/en/technical-documents/protocol/protein-biology/gel-electrophoresis/southern-and-northern-blotting)
- [Protocol for Anti Ago-RNA Immunoprecipitation from Mammalian Cells Using the RIP Kit](https://www.sigmaaldrich.com/US/en/technical-documents/protocol/protein-biology/protein-and-nucleic-acid-interactions/anti-ago-rip-from-cells)
- [Chromatin Isolation by RNA Purification (ChIRP) Protocol](https://www.sigmaaldrich.com/US/en/technical-documents/protocol/protein-biology/protein-and-nucleic-acid-interactions/chromatin-isolation-by-rna-purification)
- [Creating PLA® Probes](https://www.sigmaaldrich.com/US/en/technical-documents/protocol/protein-biology/protein-and-nucleic-acid-interactions/creating-pla-probes-duolink-application-note)
- [Using Duolink® in Protein Interaction Studies](https://www.sigmaaldrich.com/US/en/technical-documents/protocol/protein-biology/protein-and-nucleic-acid-interactions/duolink)
- [Duolink® PLA Brightfield Protocol](https://www.sigmaaldrich.com/US/en/technical-documents/protocol/protein-biology/protein-and-nucleic-acid-interactions/duolink-brightfield-user-manual)
- [Counterstaining after the Duolink *In Situ* Protocol](https://www.sigmaaldrich.com/US/en/technical-documents/protocol/protein-biology/protein-and-nucleic-acid-interactions/duolink-counterstain)
- [View More](https://www.sigmaaldrich.com/US/en/search/facet-search?focus=sitecontent&term=facet-search)
Top
__Sign In To Continue__
Sign In__Don't Have An Account?__Register
Support[Customer Support](https://www.sigmaaldrich.com/US/en/support/customer-support)[Contact Us](https://www.sigmaaldrich.com/US/en/collections/offices)[FAQ](https://maestro.my.site.com/knowledgeportal/s/)[Safety Data Sheets (SDS)](https://www.sigmaaldrich.com/US/en/documents-search?tab=sds)[Certificates (COA/COO)](https://www.sigmaaldrich.com/US/en/documents-search?tab=coa)[Quality & Regulatory](https://www.sigmaaldrich.com/US/en/life-science/quality-and-regulatory-management)[Calculators & Apps](https://www.sigmaaldrich.com/US/en/support/calculators-and-apps)[Webinars](https://www.sigmaaldrich.com/US/en/collections/webinars)
Orders[Order Lookup](https://www.sigmaaldrich.com/US/en/order-lookup)[Quick Order](https://www.sigmaaldrich.com/US/en/quick-order)[Custom Products](https://www.sigmaaldrich.com/US/en/services/custom-products)[eCommerce Solutions](https://www.sigmaaldrich.com/US/en/life-science/ecommerce/ecommerce-solutions)
Company[About Us](https://www.sigmaaldrich.com/US/en/life-science/about-us)[Responsibility](https://www.sigmaaldrich.com/US/en/life-science/ssbi)[Events](https://www.sigmaaldrich.com/US/en/collections/events)[Press Releases](https://www.sigmaaldrich.com/US/en/collections/press)[Programs](https://www.sigmaaldrich.com/US/en/life-science/partnership-programs)[Careers](https://careers.merckgroup.com/global/en)[Offices](https://www.sigmaaldrich.com/US/en/collections/offices)
Social Media
[](https://www.linkedin.com/company/merck-life-science)
[](https://x.com/Merck_lifesci)
[](https://www.facebook.com/Merck.lifescience)
[](https://www.instagram.com/mercklifescience/)
[](https://www.youtube.com/@MerckLifeScience/featured)
MilliporeSigma
__Research. Development. Production.__
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
[](https://www.sigmaaldrich.com/US/en)
[Sigma-Aldrich® Solutions](https://www.sigmaaldrich.com/US/en/life-science/sigma-aldrich)[BioReliance® Solutions](https://www.sigmaaldrich.com/US/en/life-science/bioreliance)[Millipore® Solutions](https://www.sigmaaldrich.com/US/en/life-science/millipore)[SAFC® Solutions](https://www.sigmaaldrich.com/US/en/life-science/safc)[Milli-Q® Solutions](https://www.sigmaaldrich.com/US/en/life-science/milliq)[Supelco® Solutions](https://www.sigmaaldrich.com/US/en/life-science/supelco)
© 2026 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved, including Text and Data Mining for AI training and similar technologies.
Reproduction of any materials from the site is strictly forbidden without permission.
[Site Use Terms](https://www.sigmaaldrich.com/US/en/life-science/legal/site-use-terms)|[Privacy Policy](https://www.sigmaaldrich.com/US/en/life-science/legal/privacy-statement)|[General Terms and Conditions of Sale](https://www.sigmaaldrich.com/US/en/life-science/legal/terms-and-conditions)|[Copyright Consent](https://www.sigmaaldrich.com/US/en/life-science/legal/copyright-consent) |[Site Map](https://www.sigmaaldrich.com/US/en/site-map)
An unknown error has occured.