Non-Fat Dry Milk (NFDM) Solution for Blocking Western Blots
This calculator enables the accurate preparation of this Western blot blocking solution whether you are making enough for a single experiment or multiple liters for the entire lab. Input your desired concentration and volume and click the CALCULATE button. The table will populate with the amounts of each component needed.
5% in 100 mL Recipe NFDM: % at Desired Volume: mL    
5 g of Non-fat dry milk powder g Non-fat dry milk powder
in 100 mL of 1X TBST in mL of 1X TBST
Other recipes for Western Blotting Buffers & Solutions