Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Heck Reaction in Water at Room Temperature using TPGS-750-M
Description
TPGS-750-M, DL-α-Tocopherol methoxypolyethylene glycol succinate, is a designer surfactant composed of a lipophilic α-tocopherol moiety and a hydrophilic PEG-750-M chain, joined by an inexpensive succinic acid linker. TPGS-750-M spontaneously forms micelles upon dissolution in water. The balance and composition of lipophilic and hydrophilic components have been tailored to promote a broad array of chemistry in water. Furthermore, this new, more practical surfactant can be readily be substituted for older amphiphiles, usually with equal or greater efficiency in terms of both yield and reaction times.
Conducting transition metal-catalyzed cross-coupling chemistry in water instead of organic solvent has a number of potential benefits in terms of cost, environmental impact, safety, and impurity profiles. Increasing focus on “Green” chemical processes has further promoted recent developments in this field. The actual means of implementing reactions in water, however, especially at room temperature and for water insoluble, organic substrates, has not always been clear.
One solution applied to a broad range of transition-metal-catalyzed processes is the use of small amounts of a nanomicelle-forming amphiphile in water, which provides a lipophilic medium where cross-coupling reactions can take place. Initially, the viability of surfactant promoted, transition metal-catalyzed chemistry in water at room temperature was investigated with a variety of surfactants with PTS, an α-tocopherol based diester of sebacic acid, the most effective. A number of palladium and ruthenium-catalyzed processes were found to be amenable to mild, room temperature reactions in water. Products can be recovered from the aqueous phase using standard extraction procedures and in high isolated yields.1 TPGS-750-M is a second generation surfactant designed with physical properties to promote a broader array of chemistry in water more efficiently than seen with PTS.2
Precautions
Please consult the Safety Data Sheet for information regarding hazards and safe handling practices.
Procedures
Heck Reaction – Transition-Metal-Catalyzed Cross-Coupling Reaction in Water at Room Temperature using TPGS-750-M
The catalyst Pd[P(t-Bu)3]2 (5.1 mg, 0.01 mmol, Catalog Number 676578) and aryl iodide (0.50 mmol) are added under argon into a reaction flask containing a stir bar. An aliquot of TPGS-750-M/H2O solution (1.0 mL; 5 wt % TPGS-750-M in water, Catalog Number 763918), Et3N (0.21 mL, 1.50 mmol), and acrylate/styrene (1.0 mmol) are added sequentially by syringe, and the resulting mixture is allowed to stir at room temperature for 4–12 h. The homogeneous reaction mixture is then diluted with EtOAc (2 mL) and filtered through a bed of silica gel, and the bed is further washed (3 x 5 mL) with EtOAc to collect all of the coupled material. The volatiles are removed in vacuo to afford the crude product, which is subsequently purified by flash chromatography on silica gel.
Materials
 References
  1. Lipshutz, B.H. et al., Olefin Cross-Metathesis Reactions at Room Temperature Using the Nonionic Amphiphile “PTS”: Just Add Water. Organic Letters, 10, 1325-1328 (2008).
  2. Lipshutz, B.H. et al., TPGS-750-M: A Second-Gereation Amphiphile for Metal-Catalyzed Cross-Couplings in Water at Room Temperature. J. Org. Chem., 76, 4379-4391 (2011).
Related Links