[Skip to Content](https://www.sigmaaldrich.com#main-content)
[](https://www.sigmaaldrich.com/US/en)
Products
Cart0
USEN
Products
ProductsApplicationsServicesResourcesSupport
[Login / Register](https://www.sigmaaldrich.com/oidc-sign-in)
[Order Lookup](https://www.sigmaaldrich.com/US/en/order-lookup)
[Quick Order](https://www.sigmaaldrich.com/US/en/quick-order)
Cart0
[Home](https://www.sigmaaldrich.com/US/en)[Small Molecule HPLC](https://www.sigmaaldrich.com/US/en/applications/analytical-chemistry/liquid-chromatography/small-molecule-hplc)PFAS Analysis Methods: Regulatory Alignment, QA/QC, and Contamination Control
# PFAS Analysis Methods: Regulatory Alignment, QA/QC, and Contamination Control
## Abstract
Accurate determination of Per- and polyfluoroalkyl substances (PFAS) demands precise quantification, rigorous contamination control, and validated analytical performance. This article outlines integrated approaches for reliable PFAS analysis across environmental and biological matrices, highlighting the balance between targeted LC–MS/MS workflows and broader screening techniques. Emphasis is placed on method validation, matrix-specific optimization, and harmonized global practices that ensure consistency and confidence in PFAS monitoring.
__Section Overview__
- [Why PFAS Analysis is Challenging](https://www.sigmaaldrich.com#analysis)
- [Contamination Control Essentials (Before Sampling and in the Laboratory)](https://www.sigmaaldrich.com#control)
- [QA/QC Essentials](https://www.sigmaaldrich.com#qa-qc)
- [Regulatory Landscape Snapshot](https://www.sigmaaldrich.com#regulatory)
- [Conclusion](https://www.sigmaaldrich.com#conclusion)
### At a Glance
- Choose the right method for your matrix and goal (compliance vs screening) using validated EPA/ISO/ASTM workflows.1-7
- Strong contamination control is essential at ng/L/ppt detection; avoid common PFAS sources in sampling and lab hardware, and run blanks.1,2,8
- Use isotope dilution and defined QA/QC acceptance criteria.1,2
- Align to validated regulatory workflows for defensible results.6
## [](https://www.sigmaaldrich.com)Why PFAS Analysis is Challenging
PFAS analysis poses unique analytical challenges due to their large chemical diversity and emerging/unmonitored compounds, ultra-low regulatory targets that require very high sensitivity, and ubiquitous background contamination that easily biases results.3
- __Chemical diversity:__ Thousands of PFAS have been identified, varying in chain length, functional groups (carboxylates, sulfonates, phosphates, fluorotelomers), and polarity. Analytical methods must often focus on a small subset for which reference standards are available, while suspect and non-targeted screening are required to detect emerging or unknown PFAS.2
- __Low concentration levels:__ Regulatory thresholds, such as the U.S. EPA's health advisory levels and the EU Drinking Water Directive limits, require quantitation in the low-ppt (ng/L) or even sub-ppt range. Achieving such sensitivity demands advanced instrumentation (e.g., LC–MS/MS or high-resolution MS) and meticulous contamination control.
- __Background contamination:__ PFAS are present in many laboratory materials (e.g., PTFE tubing, seals, bottle caps, solvent containers) and field sampling gear, leading to false positives if contamination is not rigorously controlled. Both field blanks and laboratory reagent blanks are mandatory in regulated PFAS methods.
- __Matrix effects:__ PFAS analysis in complex matrices such as wastewater, sludge, or biota suffers from ion suppression or enhancement during electrospray ionization. Isotope dilution with labeled analogs is the preferred approach to correct for these effects.
Given these challenges, PFAS analytical workflows must integrate contamination prevention, matrix-specific extraction, isotope dilution, and strict QA/QC as outlined in validated methods such as EPA 537.1, EPA 533, EPA 1633, SW-846 8327, the EU Drinking Water Directive (2020/2184), and Australia PFAS NEMP 3.0
| | | | | | | |
|-----------------------------------------------------|--------------------------------------|------------------------------------------------------------------------------------------------------------------------------------------------------------------------------------------------------------------------------------------------------------------------------------|-------------------------------------------------------------------------------------------------------|---------------------------------------------------------------|-------------------------------------------------------------------------------------------------------|-------------------------------------------------------------|
| Matrix/ Target | Typical Sample Volume (Guideline) | Recommended Workflow/Methods | Extraction/Cleanup | Instrument/Analysis | QC & Contamination Controls | Sources |
| Drinking water (compliance) | 250 mL–1 L | [SPE + targeted LC–MS/MS per EPA 537.1 + 533 or ISO 21675 equivalents](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/environmental-testing-and-industrial-hygiene/drinking-water-testing/lc-ms-analysis-of-pfas-compounds) | WAX or SDVB/WAX SPE → dry → elute (MeOH ± NH4OH) → concentrate | LC–MS/MS (MRM) with isotope dilution | Field/trip/equipment/lab blanks; labeled ISTDs; LCS; matrix spikes | EPA CWA PFAS methods; ISO 216751,3 |
| Drinking water (short/ultra‑short PFAS) | 250 mL–1 L | SPE tuned for short chains (mixed‑mode/WAX) + LC–MS/MS; consider HILIC | Mixed‑mode SPE or HILIC‑compatible prep | LC–MS/MS; columns that add ionic retention (mixed‑mode/HILIC) | Verify LOQs for short chains; solvent/consumable background checks | EPA \[2], ISO 21675; CIC/short-chain1,3,7 |
| Surface/ground water (non‑potable) | 250 mL–2 L | SPE (WAX/HLB) + targeted LC–MS/MS; optional AOF prescreen | SPE enrichment; ENVI‑Carb cleanup if needed | LC–MS/MS (isotope dilution) | Full blanks suite; chilled transport; avoid PTFE/FEP/PVDF/ETFE in field gear | EPA2,7,8 |
| Wastewater / high‑matrix waters | 0.1–2 L | [EPA 1633 multi‑matrix or validated SOPs; AOF prescreen where useful](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/environmental-testing-and-industrial-hygiene/waste-water-and-process-water-testing/analysis-of-pfas-in-water-according-to-epa-1633) | SLE/ASE → SPE + carbon cleanup (ENVI‑Carb) | LC–MS/MS; HRMS for suspects | Matrix spikes; surrogates; duplicates; method blanks | EPA methods (1633, 1621); TOP7,8 |
| Solids / sediments / biosolids | ~5–50 g (dry wt) | [EPA 1633 multi‑matrix or validated SOPs](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/environmental-testing-and-industrial-hygiene/waste-water-and-process-water-testing/analysis-of-pfas-in-water-according-to-epa-1633) | Dry/homogenize → ASE/Soxhlet/PLE → ENVI‑Carb + WAX/HLB SPE | LC–MS/MS (isotope dilution); HRMS as needed | Surrogates; matrix spikes; duplicates; method blanks; freeze storage | EPA methods7 |
| Food (edible commodities) | 5–50 g | [Matrix‑validated SPE/SLE/ASE; align to FDA/AOAC/GB guidance where available](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/food-and-beverage-testing-and-manufacturing/chemical-analysis-for-food-and-beverage/pfas-analysis-in-milk) | Protein precipitation (MeOH/ACN) or SLE/ASE → lipid removal (dispersive C18) → carbon cleanup | LC–MS/MS (isotope dilution); HRMS for suspects | Labeled ISTDs; matrix‑matched calibration or standard addition; method blanks; LCS; matrix spikes | FDA PFAS; AOAC PFAS China GB food/FCM9,11,12 |
| Food packaging (paper/paperboard, coatings) | Pieces or area (method‑dependent) | Screening + validated extractable PFAS methods (AOAC initiative); LC–MS/MS; 19F NMR (sum‑F) where applicable | Solvent extraction (MeOH/MeOH:ACN), sonication/ASE; SPE/SEC cleanup; hydrolysis if testing degradants | LC–MS/MS; optional 19F NMR; GC–MS for volatile precursors | Packaging/procedural blanks; surrogate recoveries; extraction efficiency checks; PFAS‑tested solvents | AOAC packaging; FDA food‑contact; China GB 4806 series11,12 |
| Food packaging migration (simulants) | 50–250 mL (per test) | Migration testing with simulants → extraction → targeted LC–MS/MS | Extract simulants (EtOH/water/acidic/oily) → SPE cleanup | LC–MS/MS; GC–MS for volatile precursors as needed | Procedural/packaging blanks; surrogate recoveries; document contact conditions | FDA food‑contact; AOAC packaging |
| Clinical / human biomonitoring (serum/plasma/urine) | Serum/plasma 0.1–1 mL; urine 1–10 mL | Protein precipitation + HybridSPE (WAX/HLB) or validated biomonitoring workflows; online SPE for throughput | Protein precipitation → SPE/CDC 6304.09; lipid cleanup for tissues | LC–MS/MS (isotope dilution); HRMS for non‑target | Labeled ISTDs; blanks; matrix spikes; recovery checks; chain‑of‑custody; PFAS‑aware handling | EPA multi‑matrix USGS/ITRC sampling QC7,8,10 |
Table 1.Overview of PFAS analytical workflows by matrix, summarizing typical sample volumes, extraction and cleanup procedures, analytical techniques, and key QA/QC measures aligned with major regulatory methods (EPA, ISO, FDA, AOAC, GB)
| | | | | | |
|---------------------------------------------------------------------------------------------------------------------------------------------------------------------------------------------------------------------------------------------------|----------------------------------------------------------------|-------------------------------------------------------|------------------------------------------------------------|-----------------------------------------------------------------------------------|-----------------------------------------------|
| Technique | What it Measures/Use Case | Typical Matrices | Strengths | Limitations/Cautions | Common Methods/Standards |
| [Targeted LC–MS/MS (SPE + LC–MS/MS)](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/environmental-testing-and-industrial-hygiene/waste-water-and-process-water-testing/analysis-of-pfas-in-water-according-to-epa-1633) | Quantifies named PFAS with high specificity | Water (drinking/non‑potable), solids, biota, clinical | Quantitative; broad analyte coverage; regulatory alignment | Requires rigorous contamination control; short‑chain retention can be challenging | EPA 537.1, 533, 1633; ISO 21675; DIN 38407‑42 |
| Direct injection LC–MS/MS (screening) | Rapid screen without SPE; confirm positives later | Non‑potable waters, some low‑matrix extracts | High throughput; simple workflow | Matrix suppression; not ideal for complex matrices | EPA 8327 |
| HRMS (suspect/non‑target) | Detects unknowns/precursors; structural clues | Any extract; research/investigative | Broader coverage; isomer info | Complex data; not routine compliance | Lab SOPs; research guidance |
| AOF (CIC) | Sum adsorbable organofluorine; prescreen burden | Water; some extracts | Total burden indicator; robust | Non‑specific; includes non‑PFAS organofluorine; requires controls | EPA 1621; CIC SOPs |
| EOF (CIC) | Sum extractable organofluorine; prescreen burden | Water, solids extracts | Total burden indicator | Non‑specific; matrix‑dependent extraction | CIC SOPs |
| TOF (CIC) | Total organofluorine; mass balance | Water; lab extracts | Mass‑balance perspective | Non‑specific; careful interpretation needed | CIC SOPs |
| TOP assay | Oxidizable precursors → PFAAs; estimates hidden precursor load | Water; solids/extracts | Reveals precursors | Semi‑quant; may miss some ethers/novel PFAS | TOP best‑practice SOPs |
| 19F NMR (sum‑19F) | Sum fluorine in extracts (including short chains like TFA) | Extracts (water/packaging/food) | Non‑specific total F check; orthogonal to CIC | Lower sensitivity vs LC–MS; needs clean extracts | Lab SOPs; method notes |
| GC–MS for volatile PFAS precursors | Volatile/semi‑volatile PFAS (e.g., FTOHs) | Air, emissions, packaging volatiles | Captures volatile precursors | Not for ionic PFAS | OTM-50 and OTM-55; lab SOPs |
Table 2.Overview of key analytical techniques used in PFAS characterization, summarizing measurement principles, typical matrices, strengths, limitations, and corresponding reference methods or standards
## [](https://www.sigmaaldrich.com)Contamination Control Essentials (Before Sampling and in the Laboratory)
- Avoid fluoropolymer contact: Sampling and analytical hardware containing PTFE, FEP, PVDF, ETFE, or related fluoropolymers can introduce trace-level contamination. Use HDPE, polypropylene, stainless steel, or glass materials when compatible.2,8
- Minimize personal sources: Wear powder-free nitrile gloves and avoid potential contamination from treated clothing, packaging materials, or personal care products.
- Implement rigorous blanks: Employ field, trip, equipment, and laboratory reagent blanks to identify and quantify background contributions; document any potential bias associated with sampling sites.1,2
- Control contamination in LC systems: Use low-bleed chromatographic columns, PFAS-tested solvents and caps, and delay or guard columns. Run system blanks routinely to monitor and correct for background levels.2
## [](https://www.sigmaaldrich.com)QA/QC Essentials
Isotope dilution: The use of isotopically labeled internal standards is standard practice to ensure quantitative accuracy and precision across analytical methods.1
Quality control samples: Apply calibration verification, laboratory control samples, matrix spikes, duplicates, and defined acceptance criteria as outlined in standard operating procedures (SOPs). Record surrogate recoveries and blank thresholds in accordance with program requirements.1,2
Reporting requirements: Ensure that method-specific limits of quantitation (LOQs) and reporting limits are achieved. Document any method modifications, such as alternative cleanup procedures, and demonstrate equivalent analytical performance.1,2,7,8
### When to use AOF/EOF/TOF/TOP and 19F NMR (Sum-Parameter Screening)
Sum-parameter approaches are applied to assess the total organofluorine or fluorine burden prior to compound-specific identification and quantification by targeted LC–MS/MS.
- __AOF (Combustion Ion Chromatography, CIC):__ Used for prescreening adsorbable organofluorine retained on activated carbon. The measurement is non-specific and may include non-PFAS compounds and therefore, targeted LC–MS/MS analysis is required for definitive speciation.
- __EOF (CIC):__ Applied for prescreening extractable organofluorine in environmental and biological matrices. Interpretation is subject to the same non-specificity caveats as AOF, and targeted LC–MS/MS should follow for compound identification.
- __TOF (CIC):__ Provides a mass-balance perspective by measuring total organofluorine content. Due to its non-specific nature, results must be interpreted cautiously and supported by targeted LC–MS/MS.
- __TOP Assay:__ Converts oxidizable precursors to their terminal perfluoroalkyl acids (PFAAs), thereby revealing hidden precursor burdens. Results are semi-quantitative and should be confirmed using targeted LC–MS/MS. High-resolution mass spectrometry (HRMS) may be applied to detect suspected or non-target PFAS species.
- __19F NMR (Sum-Fluorine):__ Quantifies total fluorine in sample extracts, including short-chain species such as trifluoroacetic acid (TFA). As a non-specific screening tool, it provides a bulk fluorine signal and must be followed by targeted LC–MS/MS for speciation and quantification.
### Decision guidance
- Unknown profile or broad screening: Start with AOF/EOF/TOF (CIC) or 19F NMR; if elevated, proceed to targeted LC–MS/MS.
- Suspected precursors: Run TOP; compare pre/post sums; confirm with targeted LC–MS/MS; consider HRMS for suspects.
- Compliance reporting: Use targeted LC–MS/MS (EPA/ISO/DIN workflows) with isotope dilution and full QA/QC; treat sum‑parameters as supporting screens.
## [](https://www.sigmaaldrich.com)Regulatory Landscape Snapshot
The monitoring and regulation of PFAS varies globally, yet several authoritative analytical methods and legislative frameworks provide the foundation for compliance testing. These approaches specify target analytes, performance criteria, and quality control measures to ensure accurate detection at ultra-trace levels.
- I__SO 21675:2019:__ International standard for determining PFAS in drinking water, surface water, and wastewater using solid-phase extraction followed by LC–MS/MS. Provides harmonized guidance on sample preparation, target analytes, and quality control requirements.
### United States (EPA)
The National Primary Drinking Water Regulation (NPDWR) has been finalized. EPA Methods 537.1 and 533 are applied for drinking water analysis. Method 1633 is used for multiple environmental matrices. Method 8327 supports direct-injection screening, and Method 1621 (AOF) is employed for organofluorine prescreening.
- EPA Method 537
LC–MS/MS method for the determination of 14 selected PFAS in drinking water, including PFOA and PFOS. Uses solid-phase extraction (SPE) for analyte concentration and cleanup, followed by negative electrospray ionization for quantitation. Designed for U.S. drinking water compliance monitoring before the expanded analyte list in 537.1 was introduced.
- [EPA Method 537.1](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/environmental-testing-and-industrial-hygiene/drinking-water-testing/lc-ms-analysis-of-pfas-compounds)
EPA Method 537.1 is designed for the determination of 18 selected PFAS in drinking water using solid-phase extraction (SPE) followed by liquid chromatography–tandem mass spectrometry (LC–MS/MS). The method targets longer-chain PFAS such as PFOS and PFOA, as well as some short-chain perfluoroalkyl acids, and is used under the Unregulated Contaminant Monitoring Rule (UCMR).
- [EPA Method 533](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/environmental-testing-and-industrial-hygiene/drinking-water-testing/lc-ms-analysis-of-pfas-compounds-in-epa-method-533)
EPA Method 533 complements 537.1 by focusing on 25 short-chain PFAS, including perfluoroalkyl acids and fluorotelomer sulfonates not addressed in Method 537.1. It employs weak anion-exchange SPE, isotope dilution, and LC–MS/MS to achieve low-ppt detection limits. Combining EPA 533 and 537.1 provides broader PFAS coverage for drinking water compliance monitoring.
- [EPA Method 1633](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/environmental-testing-and-industrial-hygiene/waste-water-and-process-water-testing/analysis-of-pfas-in-water-according-to-epa-1633)
EPA Method 1633 is the first multi-matrix PFAS method validated by the U.S. EPA and Department of Defense (DoD). It quantifies 40 PFAS in aqueous, solid, and tissue matrices using isotope dilution LC–MS/MS, with matrix-specific extraction protocols for wastewater, surface water, groundwater, soil, sediment, biosolids, and fish tissue.
- EPA Method 1621
A qualitative adsorbable organic fluorine (AOF) method for wastewater, surface water, and other aqueous matrices. Measures total [](https://www.sigmaaldrich.com)organofluorine content as a screening tool for fluorinated compounds, including PFAS, without identifying individual species. Involves adsorption onto activated carbon, combustion, and fluoride ion measurement via ion chromatography.
- [EPA Method 8327](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/environmental-testing-and-industrial-hygiene/drinking-water-testing/lc-ms-analysis-of-pfas-compounds)
EPA Method 8327 is intended for the rapid, direct injection analysis of 24 PFAS in non-potable waters without solid-phase extraction. It is primarily used for industrial discharges, groundwater, and wastewater matrices when high-throughput screening is required.
### European Union (DWD)
National implementations define specific analyte lists and limits of quantitation (LOQs). Validated LC–MS/MS workflows aligned with ISO and CEN standards are employed for targeted analysis. Sum-parameter approaches are applied where total PFAS determinations are required.
- European Union
Drinking Water Directive (2020/2184): The EU Drinking Water Directive (DWD) establishes parametric values for PFAS in water intended for human consumption. From 12 January 2026, Member States must monitor either the sum of 20 PFAS at a limit of 0.10 µg/L, or total PFAS at a limit of 0.50 µg/L. Analytical methods must achieve a limit of quantitation (LOQ) ≤ 30% of the parametric value and follow risk-based monitoring from source to tap. Laboratories are encouraged to adopt LC–MS/MS or HRMS-based methods validated for trace-level PFAS determination.
- DIN 38407-42
German national standard for quantifying selected PFAS in water using solid-phase extraction and LC–MS/MS. Serves as the methodological basis for ISO 21675 and is widely applied across European laboratories for regulatory compliance.
### Australia/NZ
The National Environmental Management Plan (NEMP) provides national guidance for PFAS monitoring and QA/QC across multiple matrices. Solid-phase extraction (SPE) followed by LC–MS/MS with isotope dilution is the recommended analytical approach.
- The PFAS National Environmental Management Plan (NEMP) 3.0
Provides a unified approach for assessing, monitoring, and managing PFAS across Australian states and territories. It includes updated guideline values for water, soil, sediments, and biota; expanded PFAS grouping approaches; and revised biosolids reuse criteria. Analytical recommendations align closely with U.S. EPA methods, emphasizing isotope dilution LC–MS/MS, strict contamination control, and matrix-specific QA/QC protocols.
### China
Standard GB 5749-2022 references PFOS and PFOA in drinking water. Broader regulation of PFAS in food contact materials is established under the GB 4806 series, including GB 4806.8-2022 for paper and paperboard. Additional limits may be introduced through local or provincial regulations.
- GB 5749-2022
Establishes national drinking water quality limits for PFAS, specifying maximum levels of 0.04 µg/L for PFOA and 0.08 µg/L for PFOS.
- [GB/T 5750.8-2023](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/environmental-testing-and-industrial-hygiene/drinking-water-testing/quantitative-analysis-of-pfas-in-drinking-water-by-lcms)
Defines the analytical method for PFAS in drinking water using SPE followed by LC–MS/MS, covering 11 target PFAS compounds at ng/L concentrations.
### Japan
A provisional combined management target of approximately 50 ng/L for PFOS + PFOA has been implemented, with progress toward formal national standards. Laboratories should follow ongoing guidance and updates from the Ministry of Health, Labour and Welfare (MHLW) and the Ministry of the Environment. (*Sources: sustainability.chemlinked*)
### Other International Guidelines and Emerging Standards
Other countries and regions have adopted or are in the process of developing PFAS monitoring and management programs. For instance, Canada has established drinking water quality guidelines for several PFAS, while Japan has set interim targets for PFOS and PFOA in surface and groundwater. In parallel, international research consortia are advancing non-targeted and total fluorine analytical approaches, which are expected to inform and shape future regulatory testing frameworks.
| | | | | |
|-----------------------------------------|--------------------------------------------------------------------|-----------------------------------------------------------------|------------------------------------------------------------|-----------------------------------------------------------------------------|
| Method | Target Matrices | Key PFAS Coverage | Analytical Technique | Primary Regulatory Use |
| EPA 537.1 | Drinking water | 18 long-chain PFAS (including PFOA, PFOS) | SPE (SDVB) + LC–MS/MS (MRM, ESI–) | U.S. EPA compliance monitoring in drinking water |
| EPA 533 | Drinking water | 25 PFAS, incl. short chain & precursors | SPE (WAX) + LC–MS/MS | U.S. EPA compliance monitoring in drinking water; short-chain PFAS coverage |
| EPA 1633 | Surface water, groundwater, wastewater, solids, biosolids, tissues | 40 PFAS | Solvent extraction + SPE clean-up + LC–MS/MS | U.S. Clean Water Act & multi-matrix regulatory programs |
| SW-846 8327 | Non-potable waters | 24 PFAS | Direct aqueous injection + LC–MS/MS | High-throughput screening; RCRA programs |
| EU Drinking Water Directive (2020/2184) | Drinking water | 20 individual PFAS (0.10 µg/L sum limit) + 0.50 µg/L total PFAS | Any validated LC–MS/MS method with LOQ ≤ 0.020 µg/L | EU drinking water compliance |
| ISO 21675:2019 | Drinking water, natural waters, wastewater | Selected PFAS (nonfiltered waters) | SPE + LC–MS/MS | International standard for PFAS in water; EU alignment |
| DIN 3840742 (F42) | Water, wastewater, sludge | Selected PFAS/PFC (commonly 10 analytes) | Solid–liquid extraction → HPLC/MSMS; WAX SPE widely used | German standard method for PFAS |
| Australia PFAS NEMP 3.0 | Water, soil, sediment, biota | Tiered PFAS list (priority & emerging) | Any validated LC–MS/MS method meeting sensitivity criteria | Australian environmental monitoring & risk assessment |
Table 3.Summary of major regulatory and standard methods for PFAS analysis, outlining target matrices, PFAS coverage, analytical techniques, and primary regulatory applications across global frameworks
## [](https://www.sigmaaldrich.com)Conclusion
PFAS analysis demands rigorous contamination control, matrix-specific extraction, and validated LC–MS/MS or HRMS techniques to achieve low-ppt detection while ensuring data integrity. Regulatory methods such as EPA 533, 537.1, and 1633, the EU Drinking Water Directive (2020/2184), and Australia’s PFAS NEMP 3.0 set strict performance and LOQ requirements, yet the analytical scope is expanding toward suspect screening, total oxidizable precursor (TOP) assays, and extractable organic fluorine (EOF) analysis. Integrating compliance-focused workflows with advanced research capabilities will be essential to address both current regulatory obligations and the evolving chemical space of PFAS contamination.
### Discover More
- [](https://www.sigmaaldrich.com)[PFAS: Definition, Classification, Sources, Impact, & Detection](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/analytical-chemistry/small-molecule-hplc/pfas-definition-classification-sources-and-impact)
- [PFAS Column & Solvent Guidance](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/analytical-chemistry/small-molecule-hplc/pfas-column-selection-for-lc-and-gc)
- [PFAS Sample Prep by Matrix](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/analytical-chemistry/small-molecule-hplc/pfas-sample-preparation-guide-by-matrix-and-method)
Explore our complete PFAS portfolio to streamline method development, meet evolving regulatory requirements, and achieve confident trace-level detection across complex matrices. Browse products, download application resources, and find the right solutions for your workflow today at [SigmaAldrich.com/PFAS](https://www.sigmaaldrich.com/US/en/campaigns/pfas-testing).
### References
1\.
EPA PFAS Drinking Water Laboratory Methods (Methods 533, 537, 537.1 overview).. Available from: [https://www.epa.gov/pfas/epa-pfas-drinking-water-laboratory-methods](https://www.epa.gov/pfas/epa-pfas-drinking-water-laboratory-methods)
2\.
EPA SW‑846 Method 8327 (direct injection LC–MS/MS for non‑potable waters). Available from: [https://www.epa.gov/system/files/documents/2021-07/8327.pdf](https://www.epa.gov/system/files/documents/2021-07/8327.pdf)
3\.
ISO 21675:2019 (Water quality — PFAS by SPE + LC–MS/MS). Available from: [https://www.iso.org/standard/71338](https://www.iso.org/standard/71338)
4\.
ASTM D7979 (PFAS in water, sludge, influent/effluent, wastewater; LC–MS/MS). Available from: [https://store.astm.org/d7979-20.html](https://store.astm.org/d7979-20.html)
5\.
ASTM D8421 (PFAS in aqueous matrices by co‑solvation + LC–MS/MS). Available from: [https://store.astm.org/d8421-22.html](https://store.astm.org/d8421-22.html)
6\.
EU Drinking Water Directive (2020/2184) — PFAS Total and Sum of PFAS parameters. Available from: [https://eur-lex.europa.eu/eli/dir/2020/2184/oj/eng](https://eur-lex.europa.eu/eli/dir/2020/2184/oj/eng)
7\.
EPA PFAS Analytical Methods Development and Sampling Research (method status incl. 1633 and 1621). Available from: [https://www.epa.gov/water-research/pfas-analytical-methods-development-and-sampling-research](https://www.epa.gov/water-research/pfas-analytical-methods-development-and-sampling-research)
8\.
USGS PFAS sampling guide (materials to avoid; blanks suite). Available from: [https://pubs.usgs.gov/publication/ofr20241001/full](https://pubs.usgs.gov/publication/ofr20241001/full)
9\.
FDA Method C‑010.03 (food/feed; QuEChERS + dSPE + SPE). Available from: [https://www.fda.gov/media/131510/download](https://www.fda.gov/media/131510/download)
10\.
ITRC PFAS Technical Guidance — Sampling and Analytical Methods. Available from: [https://pfas-1.itrcweb.org/11-sampling-and-analytical-methods/](https://pfas-1.itrcweb.org/11-sampling-and-analytical-methods/)
11\.
AOAC PFAS Initiative (foods and packaging). Available from: [https://www.aoac.org/resources/2022-pfas-initiative/](https://www.aoac.org/resources/2022-pfas-initiative/)
12\.
China GB 4806.8‑2022 (paper and paperboard in contact with foodstuffs) — summary. Available from: [https://www.qima.com/regulation/09-22/sep2022-china-published-gb-standards](https://www.qima.com/regulation/09-22/sep2022-china-published-gb-standards)
__Related Articles__
- [LC-MS/MS Analysis of 16 PFAS in Salmon Using QuEChERS Based on FDA Method C-010.02](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/food-and-beverage-testing-and-manufacturing/chemical-analysis-for-food-and-beverage/lc-ms-ms-analysis-of-16-pfas-in-salmon-using-quechers)
- [Quantitative Analysis of PFAS in Drinking Water by LCMS](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/environmental-testing-and-industrial-hygiene/drinking-water-testing/quantitative-analysis-of-pfas-in-drinking-water-by-lcms)
- [Fast and High-Resolution LC-MS Separation of PFAS](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/environmental-testing-and-industrial-hygiene/waste-water-and-process-water-testing/fast-and-high-resolution-lc-ms-separation-of-pfas)
- [Analysis of PFAS Extractables in Filtration Products Using Modified EPA 1633](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/environmental-testing-and-industrial-hygiene/waste-water-and-process-water-testing/pfas-extractables-filtration-products-using-modified-epa)
- [Analysis of PFAS Extractables in Filtration Products Using Modified EPA 537.1](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/environmental-testing-and-industrial-hygiene/waste-water-and-process-water-testing/pfas-extractables-filters-epa-537-1)
- [PFAS testing methods and guidance for sample filtration](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/environmental-testing-and-industrial-hygiene/waste-water-and-process-water-testing/pfas-testing-methods-sample-filtration)
- [Analysis of PFAS Extractables in Glass Fiber Membrane Filters After OTM-45 Front-Half Extraction](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/environmental-testing-and-industrial-hygiene/air-testing/analysis-pfas-extractables-glass-fiber-membrane-filters)
__Related Product Categories__
- [HPLC Columns](https://www.sigmaaldrich.com/US/en/products/analytical-chemistry/analytical-chromatography/hplc-columns)
- [Solid Phase Extraction (SPE) and QuEChERS](https://www.sigmaaldrich.com/US/en/products/analytical-chemistry/analytical-sample-preparation/spe-and-quechers)
- [HPLC Buffers](https://www.sigmaaldrich.com/US/en/products/analytical-chemistry/analytical-chromatography/hplc-buffers)
- [Solvents](https://www.sigmaaldrich.com/US/en/products/analytical-chemistry/analytical-chromatography/solvents)
- [Chromatography and spectroscopy reagents](https://www.sigmaaldrich.com/US/en/products/analytical-chemistry/analytical-reagents/chromatography-spectroscopy-reagents)
- [Organic Pollutant Standards](https://www.sigmaaldrich.com/US/en/products/analytical-chemistry/reference-materials/organic-pollutant-standards)
- [Certified Reference Materials](https://www.sigmaaldrich.com/US/en/products/analytical-chemistry/reference-materials/certified-reference-materials)
- [Syringe Filters](https://www.sigmaaldrich.com/US/en/products/filtration/laboratory-syringe-filters/syringe-filters)
Top
__Sign In To Continue__
Sign In__Don't Have An Account?__Register
Support[Customer Support](https://www.sigmaaldrich.com/US/en/support/customer-support)[Contact Us](https://www.sigmaaldrich.com/US/en/collections/offices)[FAQ](https://maestro.my.site.com/knowledgeportal/s/)[Safety Data Sheets (SDS)](https://www.sigmaaldrich.com/US/en/documents-search?tab=sds)[Certificates (COA/COO)](https://www.sigmaaldrich.com/US/en/documents-search?tab=coa)[Quality & Regulatory](https://www.sigmaaldrich.com/US/en/life-science/quality-and-regulatory-management)[Calculators & Apps](https://www.sigmaaldrich.com/US/en/support/calculators-and-apps)[Webinars](https://www.sigmaaldrich.com/US/en/collections/webinars)
Orders[Order Lookup](https://www.sigmaaldrich.com/US/en/order-lookup)[Quick Order](https://www.sigmaaldrich.com/US/en/quick-order)[Custom Products](https://www.sigmaaldrich.com/US/en/services/custom-products)[eCommerce Solutions](https://www.sigmaaldrich.com/US/en/life-science/ecommerce/ecommerce-solutions)
Company[About Us](https://www.sigmaaldrich.com/US/en/life-science/about-us)[Responsibility](https://www.sigmaaldrich.com/US/en/life-science/ssbi)[Events](https://www.sigmaaldrich.com/US/en/collections/events)[Press Releases](https://www.sigmaaldrich.com/US/en/collections/press)[Programs](https://www.sigmaaldrich.com/US/en/life-science/partnership-programs)[Careers](https://careers.merckgroup.com/global/en)[Offices](https://www.sigmaaldrich.com/US/en/collections/offices)
Social Media
[](https://www.linkedin.com/company/merck-life-science)
[](https://x.com/Merck_lifesci)
[](https://www.facebook.com/Merck.lifescience)
[](https://www.instagram.com/mercklifescience/)
[](https://www.youtube.com/@MerckLifeScience/featured)
MilliporeSigma
__Research. Development. Production.__
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
[](https://www.sigmaaldrich.com/US/en)
[Sigma-Aldrich® Solutions](https://www.sigmaaldrich.com/US/en/life-science/sigma-aldrich)[BioReliance® Solutions](https://www.sigmaaldrich.com/US/en/life-science/bioreliance)[Millipore® Solutions](https://www.sigmaaldrich.com/US/en/life-science/millipore)[SAFC® Solutions](https://www.sigmaaldrich.com/US/en/life-science/safc)[Milli-Q® Solutions](https://www.sigmaaldrich.com/US/en/life-science/milliq)[Supelco® Solutions](https://www.sigmaaldrich.com/US/en/life-science/supelco)
© 2026 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved, including Text and Data Mining for AI training and similar technologies.
Reproduction of any materials from the site is strictly forbidden without permission.
[Site Use Terms](https://www.sigmaaldrich.com/US/en/life-science/legal/site-use-terms)|[Privacy Policy](https://www.sigmaaldrich.com/US/en/life-science/legal/privacy-statement)|[General Terms and Conditions of Sale](https://www.sigmaaldrich.com/US/en/life-science/legal/terms-and-conditions)|[Copyright Consent](https://www.sigmaaldrich.com/US/en/life-science/legal/copyright-consent) |[Site Map](https://www.sigmaaldrich.com/US/en/site-map)