[Skip to Content](https://www.sigmaaldrich.com#main-content)
[](https://www.sigmaaldrich.com/US/en)
Products
Cart0
USEN
Products
ProductsApplicationsServicesResourcesSupport
[Login / Register](https://www.sigmaaldrich.com/oidc-sign-in)
[Order Lookup](https://www.sigmaaldrich.com/US/en/order-lookup)
[Quick Order](https://www.sigmaaldrich.com/US/en/quick-order)
Cart0
[Home](https://www.sigmaaldrich.com/US/en)[Polymerase Chain Reaction Applications](https://www.sigmaaldrich.com/US/en/applications/genomics/pcr)Easy Oligo
# Easy Oligo
[Order Now](https://www.sigmaaldrich.com/configurators/tube)
Order our Easy Oligos when the focus is speed, convenience and simplicity. Easy Oligos are shipped within 24 hours from order receipt, at a fixed concentration, and are suitable for a wide range of applications including PCR and sequencing.
__Guaranteed Yield__Minimum 3 OD (approx. 15 nmols\*)
__Format__100 µM concentration in solution
2 mL screw cap tubes
__Length__15 to 30 bases
__Purification__Fully deprotected and desalted
__Bases__DNA (A, C, G or T)
__Modifications__None
__Backbone__Phosphodiester bond
__Documentation__All oligos are delivered with Quality Assurance documentation including yields, Tm (melting temperature), MW, and µg/OD
__Quality Control__Every oligo is quality checked by mass spectrometry
__Maximum Order__48 oligos per order for 24 hour shipment
__Shipping Schedule__Ships within 24 hours of order receipt, check with your local office for ordering cut-off times
__Price__Contact your local office for pricing and availability
\*Estimate 1 OD = 5 nmols = 30 µg, for a 20 base oligo
Sorry, an unexpected error has occurred
Response not successful: Received status code 500
__Related Articles__
- [Long Oligos](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/genomics/advanced-gene-editing/custom-dna-long-oligos)
- [Custom DNA Oligos: Standard DNA Synthesis](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/genomics/pcr/standard-dna-synthesis)
- [Milli-Q® IQ 7000 Ultrapure Water System](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/analytical-chemistry/small-molecule-hplc/milli-q-iq-7000)
- [Milli-Q® IQ 7003/05/10/15 Ultrapure & Pure Lab Water Purification System](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/analytical-chemistry/wet-chemical-analysis/milli-q-iq7003-7005)
- [Custom Oligos for Commercial Use](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/genomics/iso-oligos)
- [Custom LAMP Primers in Partnership with New England Biolabs](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/genomics/pcr/custom-lamp-primers)
- [Fast siRNA](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/genomics/pcr/fast-sirna-oligos)
- [Scalable, High-Quality Custom Oligonucleotides for Commercial PCR, qPCR & NGS](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/genomics/pcr/iso-oligos)
Top
__Sign In To Continue__
Sign In__Don't Have An Account?__Register
Support[Customer Support](https://www.sigmaaldrich.com/US/en/support/customer-support)[Contact Us](https://www.sigmaaldrich.com/US/en/collections/offices)[FAQ](https://maestro.my.site.com/knowledgeportal/s/)[Safety Data Sheets (SDS)](https://www.sigmaaldrich.com/US/en/documents-search?tab=sds)[Certificates (COA/COO)](https://www.sigmaaldrich.com/US/en/documents-search?tab=coa)[Quality & Regulatory](https://www.sigmaaldrich.com/US/en/life-science/quality-and-regulatory-management)[Calculators & Apps](https://www.sigmaaldrich.com/US/en/support/calculators-and-apps)[Webinars](https://www.sigmaaldrich.com/US/en/collections/webinars)
Orders[Order Lookup](https://www.sigmaaldrich.com/US/en/order-lookup)[Quick Order](https://www.sigmaaldrich.com/US/en/quick-order)[Custom Products](https://www.sigmaaldrich.com/US/en/services/custom-products)[eCommerce Solutions](https://www.sigmaaldrich.com/US/en/life-science/ecommerce/ecommerce-solutions)
Company[About Us](https://www.sigmaaldrich.com/US/en/life-science/about-us)[Responsibility](https://www.sigmaaldrich.com/US/en/life-science/ssbi)[Events](https://www.sigmaaldrich.com/US/en/collections/events)[Press Releases](https://www.sigmaaldrich.com/US/en/collections/press)[Programs](https://www.sigmaaldrich.com/US/en/life-science/partnership-programs)[Careers](https://careers.merckgroup.com/global/en)[Offices](https://www.sigmaaldrich.com/US/en/collections/offices)
Social Media
[](https://www.linkedin.com/company/merck-life-science)
[](https://x.com/Merck_lifesci)
[](https://www.facebook.com/Merck.lifescience)
[](https://www.instagram.com/mercklifescience/)
[](https://www.youtube.com/@MerckLifeScience/featured)
MilliporeSigma
__Research. Development. Production.__
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
[](https://www.sigmaaldrich.com/US/en)
[Sigma-Aldrich® Solutions](https://www.sigmaaldrich.com/US/en/life-science/sigma-aldrich)[BioReliance® Solutions](https://www.sigmaaldrich.com/US/en/life-science/bioreliance)[Millipore® Solutions](https://www.sigmaaldrich.com/US/en/life-science/millipore)[SAFC® Solutions](https://www.sigmaaldrich.com/US/en/life-science/safc)[Milli-Q® Solutions](https://www.sigmaaldrich.com/US/en/life-science/milliq)[Supelco® Solutions](https://www.sigmaaldrich.com/US/en/life-science/supelco)
© 2026 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved, including Text and Data Mining for AI training and similar technologies.
Reproduction of any materials from the site is strictly forbidden without permission.
[Site Use Terms](https://www.sigmaaldrich.com/US/en/life-science/legal/site-use-terms)|[Privacy Policy](https://www.sigmaaldrich.com/US/en/life-science/legal/privacy-statement)|[General Terms and Conditions of Sale](https://www.sigmaaldrich.com/US/en/life-science/legal/terms-and-conditions)|[Copyright Consent](https://www.sigmaaldrich.com/US/en/life-science/legal/copyright-consent) |[Site Map](https://www.sigmaaldrich.com/US/en/site-map)
An unknown error has occured.