[Skip to Content](https://www.sigmaaldrich.com#main-content)
[](https://www.sigmaaldrich.com/US/en)
Products
Cart0
USEN
Products
ProductsApplicationsServicesResourcesSupport
[Login / Register](https://www.sigmaaldrich.com/oidc-sign-in)
[Order Lookup](https://www.sigmaaldrich.com/US/en/order-lookup)
[Quick Order](https://www.sigmaaldrich.com/US/en/quick-order)
Cart0
[Home](https://www.sigmaaldrich.com/US/en)Custom DNA Oligos: Standard DNA Synthesis
# Custom DNA Oligos: Standard DNA Synthesis
[Order Now](https://www.sigmaaldrich.com/configurators/tube)
### DNA Oligo Specifications
Synthesis Scales0.025, 0.05, 0.2, 1.0, 10, and 15µmole
[mg - gram quantity manufacturing](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/genomics/pcr/custom-iscale-oligos) available upon request
PurificationDesalt, Cartridge, HPLC, PAGE
[See Oligonucleotide Purification Guidelines](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/genomics/dna-and-rna-purification/best-purification)
LengthUp to 120 bases, consult technical support for feasibility of [longer oligos](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/genomics/advanced-gene-editing/custom-dna-long-oligos) up to 180 bases
BackbonePhosphodiester / Phosphorothioate
FormatDry or In Solution (normalized)
Modifications[Over 200 available](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/genomics/qpcr/custom-dna-oligos-modifications), including dyes (fluorescent and non-fluorescent)
Packaging2 mL - 50 mL Tubes, Microtiter Plates
Quality ControlsMALDI-TOF MS, Electrospray Ionization MS, OD by UV Spectroscopy
Technical Datasheet
& LabelsIncludes yield, Tm (melting temperature), MW, and µg/OD
Additional ServicesConcentration adjustment
Aliquoting into tubes or well plates
MSQC traces available upon request
## DNA Oligos in Plates
[Order Now (Plates)](https://www.sigmaaldrich.com/configurators/plate)
We can format your oligos to best suit your application as well as your automation and information requirements.
Custom specifications can include:
- 96 or 384 well plates: Various plate types available
- Concentration normalization options with fixed or variable volumes
- Dried or in solution (water or TE)
In addition to the common formats listed above, other options are available including:
- 1-D or 2-D Barcoding
- Customer Specified Plate and Buffers
- Daughter Plates (Copies)
- Forward & Reverse Primers in a Single Well
### Quality
Each shipment includes data sheets for import into customers’ information systems containing well location, sequence, oligo name, lot number, MW, µg/OD, length, Tm, concentration and volume (in solution).
Additional quality control measures are in place to verify accuracy for custom formats. These measures may include:
- Plate orientation and well position verification by mass spectrometry
- Mixed Primers, i.e. Forward and Reverse in a single well, verified by mass spectrometry
- Verification for machine readable barcodes (1D and 2D)
- Visual inspection of all master and daughter plates prior to shipment
[Learn more](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/genomics/pcr/custom-dna-oligos-qc-qa) about our quality control and quality assurance.
### Concentration Verification and Plate Identity
We employ a variety of liquid-handling automation systems to normalize the oligo concentration. A multi-step normalization process ensures the concentrations are within a narrow range. At the end of the process, a final absorbance reading (measured at 260 nm) ensures that every oligo is at the desired concentration.
## Custom Oligo Pools
We can provide custom oligo pools to your specification and required format. Our custom formatting capabilities have been optimized to provide complex formulations containing hundreds of oligos in a single pool.
Oligos pools can be provided normalized to a specific amount or concentration in water or buffer and supplied in liquid or dry format. Multiple aliquots of the oligo pool can be provided upon request. In addition to standard mixes, we specialize in preparing complex formulations containing varying amounts per oligo within the pool. Simple oligo mixes can be provided in plates or tubes, complex formulations containing a large number of oligos will be provided in the appropriate tube based on the pool volume or the customers required format.
Please contact [dnaoligos@milliporesigma.com](mailto:dnaoligos@milliporesigma.com) to request a quote.
### Minimum Yield Guarantees
Non-Modified Oligos (O.D./µg)*
Scale (µmole)DesaltCartridgeHPLCPAGE
0.0253/90NANANA
0.055/1501/301/300.5/15
0.212/3603/902.5/751/30
1.040/1,20012/36013/3905/150
10400/12,000NA130/3,900NA
15600/18,000NA190/5,700NA
\*Guarantees are for 20 mers or longer. Oligos shorter than 20 bases will have fewer ODs. Microgram amounts are calculated based on a 30 μg/OD value. If the μg/OD value is less than 30, the reported μg value will be proportionally less. Phosphorothioated yields are 70% of the above stated values.
### Shipping Schedule
Scale (µmole)DesaltCartridgeHPLCPAGE
0.02524 hours\*NANANA
0.051-2 days1-2 days3-4 daysInquire
0.21-2 days1-2 days3-4 daysInquire
1.01-2 days1-2 days3-4 daysInquire
Plates (non-modified)\*\*3-5 days3-5 daysInquireInquire
Greater than 1.0 µmole10 days or less10 days or less10 days or lessInquire
Large quantityInquireInquireInquireInquire
Post-synthesis modified7-10 days7-10 days7-10 daysInquire
All other modifications5-7 days or less5-7 days or less5-7 days or lessInquire
\* 24 hours is based on non-modified oligos with lengths up to 35 bases.
\** Large projects will be placed on a delivery schedule based on requirements and agreement with both parties.
Sorry, an unexpected error has occurred
Response not successful: Received status code 500
__Related Articles__
- [Scalable, High-Quality Custom Oligonucleotides for Commercial PCR, qPCR & NGS](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/genomics/pcr/iso-oligos)
- [Custom Oligos & qPCR Probes Online Tools](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/genomics/pcr/custom-oligos-and-qpcr-probes-online-tools)
- [Easy Oligo](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/genomics/pcr/easy-oligo)
- [Long Oligos](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/genomics/advanced-gene-editing/custom-dna-long-oligos)
__Related Product Categories__
- [Custom DNA Oligos & qPCR Probes](https://www.sigmaaldrich.com/US/en/products/molecular-biology-and-functional-genomics/oligos-and-qpcr-probes/custom-predesigned-dna-oligos-and-qpcr-probes)
Top
__Sign In To Continue__
Sign In__Don't Have An Account?__Register
Support[Customer Support](https://www.sigmaaldrich.com/US/en/support/customer-support)[Contact Us](https://www.sigmaaldrich.com/US/en/collections/offices)[FAQ](https://maestro.my.site.com/knowledgeportal/s/)[Safety Data Sheets (SDS)](https://www.sigmaaldrich.com/US/en/documents-search?tab=sds)[Certificates (COA/COO)](https://www.sigmaaldrich.com/US/en/documents-search?tab=coa)[Quality & Regulatory](https://www.sigmaaldrich.com/US/en/life-science/quality-and-regulatory-management)[Calculators & Apps](https://www.sigmaaldrich.com/US/en/support/calculators-and-apps)[Webinars](https://www.sigmaaldrich.com/US/en/collections/webinars)
Orders[Order Lookup](https://www.sigmaaldrich.com/US/en/order-lookup)[Quick Order](https://www.sigmaaldrich.com/US/en/quick-order)[Custom Products](https://www.sigmaaldrich.com/US/en/services/custom-products)[eCommerce Solutions](https://www.sigmaaldrich.com/US/en/life-science/ecommerce/ecommerce-solutions)
Company[About Us](https://www.sigmaaldrich.com/US/en/life-science/about-us)[Responsibility](https://www.sigmaaldrich.com/US/en/life-science/ssbi)[Events](https://www.sigmaaldrich.com/US/en/collections/events)[Press Releases](https://www.sigmaaldrich.com/US/en/collections/press)[Programs](https://www.sigmaaldrich.com/US/en/life-science/partnership-programs)[Careers](https://careers.merckgroup.com/global/en)[Offices](https://www.sigmaaldrich.com/US/en/collections/offices)
Social Media
[](https://www.linkedin.com/company/merck-life-science)
[](https://x.com/Merck_lifesci)
[](https://www.facebook.com/Merck.lifescience)
[](https://www.instagram.com/mercklifescience/)
[](https://www.youtube.com/@MerckLifeScience/featured)
MilliporeSigma
__Research. Development. Production.__
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
[](https://www.sigmaaldrich.com/US/en)
[Sigma-Aldrich® Solutions](https://www.sigmaaldrich.com/US/en/life-science/sigma-aldrich)[BioReliance® Solutions](https://www.sigmaaldrich.com/US/en/life-science/bioreliance)[Millipore® Solutions](https://www.sigmaaldrich.com/US/en/life-science/millipore)[SAFC® Solutions](https://www.sigmaaldrich.com/US/en/life-science/safc)[Milli-Q® Solutions](https://www.sigmaaldrich.com/US/en/life-science/milliq)[Supelco® Solutions](https://www.sigmaaldrich.com/US/en/life-science/supelco)
© 2026 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved, including Text and Data Mining for AI training and similar technologies.
Reproduction of any materials from the site is strictly forbidden without permission.
[Site Use Terms](https://www.sigmaaldrich.com/US/en/life-science/legal/site-use-terms)|[Privacy Policy](https://www.sigmaaldrich.com/US/en/life-science/legal/privacy-statement)|[General Terms and Conditions of Sale](https://www.sigmaaldrich.com/US/en/life-science/legal/terms-and-conditions)|[Copyright Consent](https://www.sigmaaldrich.com/US/en/life-science/legal/copyright-consent) |[Site Map](https://www.sigmaaldrich.com/US/en/site-map)