[Skip to Content](https://www.sigmaaldrich.com#main-content) [![MilliporeSigma](https://www.sigmaaldrich.com/static/logos/purple/millipore_sigma.svg)](https://www.sigmaaldrich.com/US/en) Products Cart0 USEN Products ProductsApplicationsServicesResourcesSupport [Login / Register](https://www.sigmaaldrich.com/oidc-sign-in) [Order Lookup](https://www.sigmaaldrich.com/US/en/order-lookup) [Quick Order](https://www.sigmaaldrich.com/US/en/quick-order) Cart0 [Home](https://www.sigmaaldrich.com/US/en)[Polymerase Chain Reaction Applications](https://www.sigmaaldrich.com/US/en/applications/genomics/pcr)WellRED Oligos # WellRED Oligos [Order Now](https://www.sigmaaldrich.com/configurators/tube) Designed for use with the CEQ/GeXP Genetic Analysis Systems, we are pleased to offer WellRED dye-labeled oligos under our license from Beckman Coulter, Inc. In addtion to their designed application, these labeled oligos may also be used in direct hybridization or PCR amplification. WellRED dye-labeled oligos are DNA oligonucleotides labeled with WellRED dyes, D2, D3 or D4, at the 5'end. ### WellRED Specifiations YieldsCartridge: 2 and 4 OD HPLC: 5, 10, 50 and 100 OD PurificationCartridge, HPLC Length15 – 30 bases (inquire for longer lengths) BackbonePhosphodiester FormatSupplied Dry ModificationsD2-PA, D3-PA or D4-PA at the 5’end Packaging2 mL Opaque Tubes Quality ControlsMALDI-TOF MS, Electrospray Ionization MS, OD by UV Spectroscopy Technical Datasheet & LabelsIncludes yield, Tm (melting temperature), MW, and µg/OD Duplicate set of labels with each order Additional ServicesAliquoting into tubes MSQC traces available upon request Shipping ScheduleShipped within 4 to 5 business days ## Benefits of WellRED Dye Chemistry - WellRED oligos use cyanine-based fluorescent dyes with high extinction coefficients that absorb in the near-infrared region, thereby reducing background noise from biological materials, resulting in highly sensitive detection. - The CEQ/GeXP Genetic Analysis Systems enable DNA samples to be kept in linear form during analysis. The CEQ/GeXP Genetic Analysis Systems' online denaturation process prevents any reformation of secondary structures, especially during a long run without any manual steps. - WellRED dye chemistry and pre-heating enable the CEQ/GeXP Genetic Analysis Systems to resolve complex structures, where other systems fail. ### Yields and Estimated Number of Assays Yield (OD)24510 PurificationCartridgeCartridgeHPLCHPLC Approx. yield (nmols\*)10202550 Estimated assays (reactions)1,000-2,0002,000-4,0002,500-5,0005,000-10,000 \*Estimate 1 OD = 5 nmols = 30 µg, for a 20 base oligo ### Spectral Properties of WellRED Dyes WellRED OligoExcitation maximum (nm)Emission maximum (nm)Molar extinction coefficient D2-PA750770170,000 D3-PA685706224,000 D4-PA650670203,000 Sorry, an unexpected error has occurred Response not successful: Received status code 500 Top __Sign In To Continue__ To continue reading please sign in or create an account. Sign In__Don't Have An Account?__Register Support[Customer Support](https://www.sigmaaldrich.com/US/en/support/customer-support)[Contact Us](https://www.sigmaaldrich.com/US/en/collections/offices)[FAQ](https://maestro.my.site.com/knowledgeportal/s/)[Safety Data Sheets (SDS)](https://www.sigmaaldrich.com/US/en/documents-search?tab=sds)[Certificates (COA/COO)](https://www.sigmaaldrich.com/US/en/documents-search?tab=coa)[Quality & Regulatory](https://www.sigmaaldrich.com/US/en/life-science/quality-and-regulatory-management)[Calculators & Apps](https://www.sigmaaldrich.com/US/en/support/calculators-and-apps)[Webinars](https://www.sigmaaldrich.com/US/en/collections/webinars) Orders[Order Lookup](https://www.sigmaaldrich.com/US/en/order-lookup)[Quick Order](https://www.sigmaaldrich.com/US/en/quick-order)[Custom Products](https://www.sigmaaldrich.com/US/en/services/custom-products)[eCommerce Solutions](https://www.sigmaaldrich.com/US/en/life-science/ecommerce/ecommerce-solutions) Company[About Us](https://www.sigmaaldrich.com/US/en/life-science/about-us)[Responsibility](https://www.sigmaaldrich.com/US/en/life-science/ssbi)[Events](https://www.sigmaaldrich.com/US/en/collections/events)[Press Releases](https://www.sigmaaldrich.com/US/en/collections/press)[Programs](https://www.sigmaaldrich.com/US/en/life-science/partnership-programs)[Careers](https://careers.merckgroup.com/global/en)[Offices](https://www.sigmaaldrich.com/US/en/collections/offices) Social Media [![LinkedIn icon](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/icons/linkedin.svg)](https://www.linkedin.com/company/merck-life-science) [![X icon](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/icons/x-icon.svg)](https://x.com/Merck_lifesci) [![Facebook Icon](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/icons/facebook.svg)](https://www.facebook.com/Merck.lifescience) [![Instagram Icon](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/icons/instagram.svg)](https://www.instagram.com/mercklifescience/) [![YouTube Icon](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/icons/youtube.svg)](https://www.youtube.com/@MerckLifeScience/featured) MilliporeSigma __Research. Development. Production.__ We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production. [![Vibrant M string](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/icons/vibrant-m-string-1-rrvcrp.svg)](https://www.sigmaaldrich.com/US/en) [Sigma-Aldrich® Solutions](https://www.sigmaaldrich.com/US/en/life-science/sigma-aldrich)[BioReliance® Solutions](https://www.sigmaaldrich.com/US/en/life-science/bioreliance)[Millipore® Solutions](https://www.sigmaaldrich.com/US/en/life-science/millipore)[SAFC® Solutions](https://www.sigmaaldrich.com/US/en/life-science/safc)[Milli-Q® Solutions](https://www.sigmaaldrich.com/US/en/life-science/milliq)[Supelco® Solutions](https://www.sigmaaldrich.com/US/en/life-science/supelco) © 2026 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved, including Text and Data Mining for AI training and similar technologies. Reproduction of any materials from the site is strictly forbidden without permission. [Site Use Terms](https://www.sigmaaldrich.com/US/en/life-science/legal/site-use-terms)|[Privacy Policy](https://www.sigmaaldrich.com/US/en/life-science/legal/privacy-statement)|[General Terms and Conditions of Sale](https://www.sigmaaldrich.com/US/en/life-science/legal/terms-and-conditions)|[Copyright Consent](https://www.sigmaaldrich.com/US/en/life-science/legal/copyright-consent) |[Site Map](https://www.sigmaaldrich.com/US/en/site-map) An unknown error has occured.