[Skip to Content](https://www.sigmaaldrich.com#main-content)
[](https://www.sigmaaldrich.com/US/en)
Products
Cart0
USEN
Products
ProductsApplicationsServicesResourcesSupport
[Login / Register](https://www.sigmaaldrich.com/oidc-sign-in)
[Order Lookup](https://www.sigmaaldrich.com/US/en/order-lookup)
[Quick Order](https://www.sigmaaldrich.com/US/en/quick-order)
Cart0
[Home](https://www.sigmaaldrich.com/US/en)[qPCR](https://www.sigmaaldrich.com/US/en/applications/genomics/qpcr)Custom Oligos Modifications
# Custom Oligos Modifications
We offer over 200 modifications for both DNA and RNA oligonucleotides and probes. We can attach fluorescent dyes to oligos at the 5'-end, 3'-end and internal positions. Our product offering includes common dye modifiers such as 6-FAM™, HEX™, and TET™, as well as exotic modifications.
In addition to standard fluorescent modifications, we have the capabilities to provide [D](https://www.sigmaaldrich.com/US/en/custom-pdp/de44feb2-4e6f-47f8-9549-adff8bf4a39c)[ual-Labeled Probes](https://www.sigmaaldrich.com/US/en/custom-pdp/de44feb2-4e6f-47f8-9549-adff8bf4a39c) including Molecular Beacons, LightCycler® Probes, and Scorpions® Probes.
- [Minimum Yield Guarantees](https://www.sigmaaldrich.com#yield)
- [Shipping Schedule](https://www.sigmaaldrich.com#shipping)
- [Quality Control](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/genomics/pcr/custom-dna-oligos-qc-qa)
Modification Categories
- [Fluorescent](https://www.sigmaaldrich.com#fluorescent)
- [Attachment](https://www.sigmaaldrich.com#attachment)
- [Binding](https://www.sigmaaldrich.com#binding)
- [Spacer](https://www.sigmaaldrich.com#spacer)
- [Analog](https://www.sigmaaldrich.com#analog)
- [Intercalation](https://www.sigmaaldrich.com#intercalation)
- [Antisense](https://www.sigmaaldrich.com#antisense)
- [Phosphorothioates](https://www.sigmaaldrich.com#phosphorothioates)
[Modification Technical Overview (PDF)](https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/390/354/custom-oligos-modifications-ms.pdf)
[](https://www.sigmaaldrich.com)FluorescentPositionsExcitation
Maximum (nm)Emission
Maximum (nm)Fluorescent Color
6-FAM™ (fluorescein)5' End, 3' End495520Green
ROX™5' End, 3' End586610Orange
Cyanine 35' End, 3' End549566Green-Yellow
Cyanine 55' End, 3' End646669Red
Cyanine 5.55' End675694Red
6-FAM dT5' End, Internal495520Green
HEX™5' End535556Green
JOE™5' End, 3' End529555Green
6-Carboxy-rhodamine 6G™5' End, 3' End524550Green
TAMRA3' End557583Green-Yellow
TAMRA NHS Ester5' End557583Yellow
TET™5' End521536Green
TxRd (Sulforhodamine 101-X)5' End, 3' End597616Red-Orange
A488 (Sulfonated Fluorescein 488)5' End, 3' End495519Green
[WellRED D2-PA](https://www.sigmaaldrich.com/US/en/custom-pdp/f2caf8b5-d59f-4814-beed-aaea667a16ab)5' End750770Red-Infrared
[WellRED D3-PA](https://www.sigmaaldrich.com/US/en/custom-pdp/f2caf8b5-d59f-4814-beed-aaea667a16ab)5' End685706Red
[WellRED D4-PA](https://www.sigmaaldrich.com/US/en/custom-pdp/f2caf8b5-d59f-4814-beed-aaea667a16ab)5' End650670Red
[](https://www.sigmaaldrich.com)AttachmentPositionsPurifications
__Amine__
DMS(O)MT-Amino-Modifier-C65' EndCartridge, HPLC
Amino-Modifier-C3-TFA5' EndDesalt, HPLC
Amino-Modifier-C125' EndCartridge, HPLC
Amino-Modifier-C6-TFA5' EndDesalt, PAGE
Amino-dT5' EndCartridge, HPLC
Amino-Modifier-55' EndCartridge, HPLC
Amino-Modifier-C2-dTInternalDesalt, Cartridge, HPLC, PAGE
Amino-Modifier-C6-dTInternalDesalt, Cartridge, HPLC, PAGE
3'-Amino-Modifier-C73' EndDesalt, Cartridge, HPLC, PAGE
__Glycerol__
Glyceryl3' EndDesalt, Cartridge, HPLC, PAGE
__Phosphate__
Phosphate3', 5' EndDesalt, PAGE (5')
Desalt, Cartridge, HPLC, PAGE (3')
Phosphate II
5' EndCartridge, HPLC
__Thiol__
5'-Thiol-Modifier C6 S-S5' End, InternalCartridge, HPLC
3'-Thiol-Modifier-C3 S-S3' EndDesalt, Cartridge, HPLC, PAGE
[](https://www.sigmaaldrich.com)BindingPositionsPurifications
__Biotin__
5'-Biotin5' EndDesalt, Cartridge, HPLC, PAGE
BiotinInternalHPLC, PAGE
Biotin-dT5' End, InternalHPLC, PAGE
Biotin-TEG5' End, InternalCartridge, HPLC
3'-Biotin-TEG3' EndHPLC, PAGE
__Digoxigenin__
Digoxigenin5' End, 3' End
HPLC
__Dinitrophenol__
2,4-Dinitrophenyl-TEG5' End, InternalHPLC
[](https://www.sigmaaldrich.com)SpacerPositionsPurifications
__Spacer__
Spacer 95' End, InternalDesalt, Cartridge, HPLC, PAGE
Spacer 125' End, InternalDesalt, Cartridge, HPLC, PAGE
Spacer 185' End, InternalDesalt, Cartridge, HPLC, PAGE
Spacer C35' End, InternalDesalt, Cartridge, HPLC, PAGE
3'-Spacer-C33' EndDesalt, Cartridge, HPLC, PAGE
[](https://www.sigmaaldrich.com)AnalogPositionsPurifications
__Chain Terminating__
3'-dA3' EndDesalt, Cartridge, HPLC, PAGE
3'-dG3' EndDesalt, Cartridge, HPLC, PAGE
3'-dC3' EndDesalt, Cartridge, HPLC, PAGE
3'-dT3' EndDesalt, Cartridge, HPLC, PAGE
__Chemotherapeutic__
Cytosine Arabinoside (araC)InternalDesalt, Cartridge, HPLC, PAGE
2',3'-dideoxycytidine (ddC)3' EndDesalt, Cartridge, HPLC, PAGE
__Contamination Prevention__
Ribo U3' EndDesalt, Cartridge, HPLC, PAGE
__Crosslinking__
8-Br-dA5' End, InternalDesalt, Cartridge, HPLC, PAGE
8-Br-dG5' End, InternalDesalt, Cartridge, HPLC, PAGE
5-Br-dC5' End, InternalDesalt, Cartridge, HPLC, PAGE
5-Br-dU5' End, InternalDesalt, Cartridge, HPLC, PAGE
5-F-dU5' End, InternalDesalt, Cartridge, HPLC, PAGE
5-I-dC5' End, InternalDesalt, Cartridge, HPLC, PAGE
5-I-dU5' End, InternalDesalt, Cartridge, HPLC, PAGE
__Damage and Repair__
2'-Deoxypseudouridine5' End, InternalDesalt, Cartridge, HPLC, PAGE
5,6-Dihydro-dT5' End, InternalDesalt, Cartridge, HPLC, PAGE
5,6-Dihydro-dU5' End, InternalDesalt, Cartridge, HPLC, PAGE
5-OH-dC5' End, InternalDesalt, Cartridge, HPLC, PAGE
5-OH-dU5' End, InternalDesalt, Cartridge, HPLC, PAGE
8-Oxo-dA5' End, InternalDesalt, Cartridge, HPLC, PAGE
8-Oxo-dG5' End, InternalDesalt, Cartridge, HPLC, PAGE
Thymidine Glycol5' End, InternalPAGE
TMP-F-dU, F-dCInternalDesalt, Cartridge, HPLC, PAGE
dUracil5' End, Internal,
3' EndDesalt, Cartridge, HPLC, PAGE
__Degenerate__
2'-Deoxynebularine5' End, InternalCartridge, HPLC, PAGE
Derivative K (dK)5' End, Internal,
3' EndDesalt, Cartridge, HPLC, PAGE
Derivative P (dP)5' End, Internal,
3' EndDesalt, Cartridge, HPLC, PAGE
Inosine5' End, Internal,
3' EndDesalt, Cartridge, HPLC, PAGE
5-Nitroindole5' End, Internal,
5' EndDesalt, Cartridge, HPLC, PAGE
3-Nitropyrrole5' End, Internal,
3' EndDesalt (Internal only), Cartridge, HPLC, PAGE
__Duplex Stabilizing__
2,6-Diaminopurine5' End, InternalDesalt, Cartridge, HPLC, PAGE
5-Me-dC5' End, InternalDesalt, Cartridge, HPLC, PAGE
__Fluorescent__
2-Aminopurine5' End, InternalDesalt, Cartridge, HPLC, PAGE
Etheno-dA5' End, InternalDesalt, Cartridge, HPLC, PAGE
__Hybridization Specificity__
[Locked Nucleic Acid](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/genomics/pcr/locked-nucleic-acids-faq)5' End, Internal,
3' EndCartridge, HPLC, PAGE
__Methylation Mutagenesis__
N6-Me-dA5' End, InternalDesalt, Cartridge, HPLC, PAGE
O6-Me-dG5' End, InternalDesalt, Cartridge, HPLC, PAGE
O4-Me-dT5' End, InternalDesalt, Cartridge, HPLC, PAGE
__No Base__
dSpacerInternalDesalt, Cartridge, HPLC, PAGE
__siRNA Guide Strand Selection__
5'-OMe-dT5' EndDesalt, HPLC, PAGE
__Structure and Function__
7-Deaza-dA5' End, InternalDesalt, Cartridge, HPLC, PAGE
7-Deaza-dG5' End, InternalDesalt, Cartridge, HPLC, PAGE
7-Deaza-dX5' End, InternalDesalt, Cartridge, HPLC, PAGE
7-Deaza-8-Aza-dA5' End, InternalDesalt, Cartridge, HPLC, PAGE
dA-5' (Inverse Oligo)5' End, Internal,
3' EndDesalt, Cartridge, HPLC, PAGE
dG-5' (Inverse Oligo)5' End, Internal,
3' EndDesalt, Cartridge, HPLC, PAGE
dC-5' (Inverse Oligo)5' End, Internal,
3' EndDesalt, Cartridge, HPLC, PAGE
dT-5' (Inverse Oligo)5' End, Internal,
3' EndDesalt, Cartridge, HPLC, PAGE
__Translation Termination__
Puromycin3' EndDesalt, Cartridge, HPLC, PAGE
[](https://www.sigmaaldrich.com)IntercalationPositionsPurifications
__Acridine__
AcridineInternalDesalt, Cartridge, HPLC, PAGE
__Psoralen__
C25' EndDesalt, Cartridge, HPLC, PAGE
C65' EndDesalt, Cartridge, HPLC, PAGE
[](https://www.sigmaaldrich.com)AntisensePositionsPurifications
__Cholesterol__
Cholesteryl-TEG5' End, InternalCartridge, HPLC
3'-Cholesteryl-TEG3' EndDesalt, Cartridge, HPLC, PAGE
__Methyl RNA__
2'-OMe-RNA A5' End, Internal,
3' EndDesalt, Cartridge, HPLC, PAGE
2'-OMe-RNA G5' End, Internal,
3' EndDesalt, Cartridge, HPLC, PAGE
2'-OMe-RNA C5' End, Internal,
3' EndDesalt, Cartridge, HPLC, PAGE
2'-OMe-RNA U5' End, Internal,
3' EndDesalt, Cartridge, HPLC, PAGE
__Methoxyethyl RNA__
2'-MOE-RNA A5' End, Internal,
3' EndDesalt, Cartridge, HPLC, PAGE
2'-MOE-RNA G5' End, Internal,
3' EndDesalt, Cartridge, HPLC, PAGE
2'-MOE-RNA C5' End, Internal,
3' EndDesalt, Cartridge, HPLC, PAGE
2'-MOE-RNA U5' End, Internal,
3' EndDesalt, Cartridge, HPLC, PAGE
__Fluoro RNA__
2'-F-RNA A5' End, Internal,
3' EndDesalt, Cartridge, HPLC, PAGE
2'-F-RNA G5' End, Internal,
3' EndDesalt, Cartridge, HPLC, PAGE
2'-F-RNA C5' End, Internal,
3' EndDesalt, Cartridge, HPLC, PAGE
2'-F-RNA U5' End, Internal,
3' EndDesalt, Cartridge, HPLC, PAGE
__Thiophosphate__
Thiophosphate (S-Oligo)5' End, Internal,
3' EndDesalt, Cartridge (5' End, Internal)
Desalt, Cartridge, HPLC, PAGE (3' End)
__2'__ → __5' Synthesis__
dA-5'5' End, Internal,
3' EndDesalt, Cartridge, HPLC, PAGE
dG-5'5' End, Internal,
3' EndDesalt, Cartridge, HPLC, PAGE
dC-5'5' End, Internal,
3' EndDesalt, Cartridge, HPLC, PAGE
dT-5'5' End, Internal,
3' EndDesalt, Cartridge, HPLC, PAGE
## Phosphorothioates (or S-oligos)
[Phosphorothioates (or S-oligos)](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/genomics/gene-expression-and-silencing/phosphorothioates) are a variant of normal DNA in which one of the non-bridging oxygens is replaced by sulfur. The sulfurization of the internucleotide bond creates a molecule that is resistant to cellular nuclease degradation. Sulfur bonds can be introduced at the 5’, 3’, or internal positions.
[](https://www.sigmaaldrich.com)
### Minimum Yield Guarantees
__Modified Oligos (O.D./µg)\*__
__Scale (µmole)____Desalt____Cartridge____HPLC____PAGE__
0.052/600.4/120.4/120.2/6
0.25/1501/301/300.4/12
1.016/4805/1505/1502/60
10160/4,800NA52/1,560NA
15240/7,200NA76/2,280NA
*\*Guarantees are for 20 mers or longer. Oligos shorter than 20 bases will have fewer ODs. Microgram amounts are calculated based on a 30 μg/OD value. If the μg/OD value is less than 30, the reported μg value will be proportionally less. Phosphorothioated yields are 70% of the above stated values. Post-synthesis modifications may yield 50% less than the above stated values.*
| | | | | |
|------------------------------------------------------------|------------|---------------|-----------|-----------|
| __Non-Modified Phosphorothioates (S-Oligos) (O.D./µg )\*__ | | | | |
| __Scale (µmole)__ | __Desalt__ | __Cartridge__ | __HPLC__ | __PAGE__ |
| 0.05 | 3.5/105 | 0.7/21 | 0.7/21 | 0.35/10.5 |
| 0.2 | 8.4/252 | 2.1/63 | 1.75/52.5 | 0.7/21 |
| 1.0 | 28/840 | 8.4/252 | 9.1/273 | 3.5/105 |
| 10 | 280/8,400 | NA | 91/2,730 | NA |
| 15 | 420/12,600 | NA | 133/3,990 | NA |
\*Guarantees are for 20 mers or longer. Oligos shorter than 20 bases will have fewer ODs. Microgram amounts are calculated based on a 30 μg/OD value. If the μg/OD value is less than 30, the reported μg value will be proportionally less.
[](https://www.sigmaaldrich.com)
### Shipping Schedule
__Purification____Desalt____Cartridge____HPLC____PAGE__
Standard Modifications5-7 days or less5-7 days or less5-7 days or lessInquire
Post-Synthesis Modifications7-10 days7-10 days7-10 daysInquire
\*For complete label license information, please visit [sigmaaldrich.com/oligolicenses](https://www.sigmaaldrich.com/technical-documents/articles/biology/label-licenses.html).
[](https://www.sigmaaldrich.com)
## Materials
Sorry, an unexpected error has occurred
Response not successful: Received status code 500
__Related Articles__
- [PCR/qPCR/dPCR Assay Design](https://www.sigmaaldrich.com/US/en/technical-documents/protocol/genomics/pcr/pcr-qpcr-dpcr-assay-design)
- [Cas9-Mediated Recombineering for Kdo2-Lipid A](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/genomics/advanced-gene-editing/crispr-cas-systems)
- [Mgat4 May Play a Role in Increased Sialylation by Overexpressing Functional MGAT1 in Mgat1-Disrupted Chinese Hamster Ovary (CHO) Cells](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/genomics/cloning-and-expression/mgat1-disrupted-chinese-hamster-ovary-cells)
- [Plasmid Product Nomenclature](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/genomics/cloning-and-expression/plasmid-nomenclature)
- [Impact of GenElute™-E Purification on DNA Processes](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/genomics/dna-and-rna-purification/dna-purification-methods)
- [Extract-N-Amp™ Plant PCR Kits](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/genomics/dna-and-rna-purification/extract-n-amp-plant-kits)
- [Complete Solutions for PCR Assay Development](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/genomics/dna-and-rna-purification/pcr-assay-development)
- [Genomic DNA Preparation Troubleshooting](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/genomics/dna-and-rna-purification/problems-during-genomic-dna-preparation)
- [View More](https://www.sigmaaldrich.com/US/en/search/facet-search?focus=sitecontent&term=facet-search)
__Related Product Categories__
- [Custom DNA Oligos & qPCR Probes](https://www.sigmaaldrich.com/US/en/products/molecular-biology-and-functional-genomics/oligos-and-qpcr-probes/custom-predesigned-dna-oligos-and-qpcr-probes)
Top
__Sign In To Continue__
Sign In__Don't Have An Account?__Register
Support[Customer Support](https://www.sigmaaldrich.com/US/en/support/customer-support)[Contact Us](https://www.sigmaaldrich.com/US/en/collections/offices)[FAQ](https://maestro.my.site.com/knowledgeportal/s/)[Safety Data Sheets (SDS)](https://www.sigmaaldrich.com/US/en/documents-search?tab=sds)[Certificates (COA/COO)](https://www.sigmaaldrich.com/US/en/documents-search?tab=coa)[Quality & Regulatory](https://www.sigmaaldrich.com/US/en/life-science/quality-and-regulatory-management)[Calculators & Apps](https://www.sigmaaldrich.com/US/en/support/calculators-and-apps)[Webinars](https://www.sigmaaldrich.com/US/en/collections/webinars)
Orders[Order Lookup](https://www.sigmaaldrich.com/US/en/order-lookup)[Quick Order](https://www.sigmaaldrich.com/US/en/quick-order)[Custom Products](https://www.sigmaaldrich.com/US/en/services/custom-products)[eCommerce Solutions](https://www.sigmaaldrich.com/US/en/life-science/ecommerce/ecommerce-solutions)
Company[About Us](https://www.sigmaaldrich.com/US/en/life-science/about-us)[Responsibility](https://www.sigmaaldrich.com/US/en/life-science/ssbi)[Events](https://www.sigmaaldrich.com/US/en/collections/events)[Press Releases](https://www.sigmaaldrich.com/US/en/collections/press)[Programs](https://www.sigmaaldrich.com/US/en/life-science/partnership-programs)[Careers](https://careers.merckgroup.com/global/en)[Offices](https://www.sigmaaldrich.com/US/en/collections/offices)
Social Media
[](https://www.linkedin.com/company/merck-life-science)
[](https://x.com/Merck_lifesci)
[](https://www.facebook.com/Merck.lifescience)
[](https://www.instagram.com/mercklifescience/)
[](https://www.youtube.com/@MerckLifeScience/featured)
MilliporeSigma
__Research. Development. Production.__
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
[](https://www.sigmaaldrich.com/US/en)
[Sigma-Aldrich® Solutions](https://www.sigmaaldrich.com/US/en/life-science/sigma-aldrich)[BioReliance® Solutions](https://www.sigmaaldrich.com/US/en/life-science/bioreliance)[Millipore® Solutions](https://www.sigmaaldrich.com/US/en/life-science/millipore)[SAFC® Solutions](https://www.sigmaaldrich.com/US/en/life-science/safc)[Milli-Q® Solutions](https://www.sigmaaldrich.com/US/en/life-science/milliq)[Supelco® Solutions](https://www.sigmaaldrich.com/US/en/life-science/supelco)
© 2026 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved, including Text and Data Mining for AI training and similar technologies.
Reproduction of any materials from the site is strictly forbidden without permission.
[Site Use Terms](https://www.sigmaaldrich.com/US/en/life-science/legal/site-use-terms)|[Privacy Policy](https://www.sigmaaldrich.com/US/en/life-science/legal/privacy-statement)|[General Terms and Conditions of Sale](https://www.sigmaaldrich.com/US/en/life-science/legal/terms-and-conditions)|[Copyright Consent](https://www.sigmaaldrich.com/US/en/life-science/legal/copyright-consent) |[Site Map](https://www.sigmaaldrich.com/US/en/site-map)