[Skip to Content](https://www.sigmaaldrich.com#main-content)
[](https://www.sigmaaldrich.com/US/en)
Products
Cart0
USEN
Products
ProductsApplicationsServicesResourcesSupport
[Login / Register](https://www.sigmaaldrich.com/oidc-sign-in)
[Order Lookup](https://www.sigmaaldrich.com/US/en/order-lookup)
[Quick Order](https://www.sigmaaldrich.com/US/en/quick-order)
Cart0
[Home](https://www.sigmaaldrich.com/US/en)[Sequencing](https://www.sigmaaldrich.com/US/en/applications/genomics/sequencing)Amino Acid Codon Wheel
# Amino Acid Codon Wheel
An amino acid codon wheel (also known as an amino acid color wheel) is a useful tool to find which amino acid is translated from your RNA sequence. Codon wheels are used by scientists, researchers and students during RNA translation to find the amino acids for that sequence as a quick, easy reference tool.
## How to Use an Amino Acid Wheel
To use an amino acid codon wheel, start from the center and follow the RNA codons until you have the 3 nucleotide bases. Next, translate the three bases into an amino acid from the mRNA codons. The process is called RNA translation. Once established, follow the RNA sequence to find the amino acid that it translates to. The protein that mRNA translates to is made up of those amino acids.
Use this amino acid codon wheel during RNA translation to discover the amino acids related to your sequence.
## Download & Print Amino Acid Wheels
These files are available to download for your own printable amino acid codon wheel.
Downloadable Amino Acid Wheels in [JPG](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/technical-documents/articles/genomics/sequencing/amino-acids-codon-wheel.jpg) or [PNG](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/technical-documents/articles/genomics/sequencing/amino-acids-codon-wheel.png) formats. Or, launch the [amino acid codon wheel SVG](https://www.sigmaaldrich.com/content/dam/cms-commons/sigmaaldrich/marketing/global/images/technical-documents/articles/genomics/sequencing/amino-acids-codon-wheel.svg) in a new browser window.
This content is provided free and available to re-use with proper link back to the resource and mention of our domain. Please bookmark and share this page.
## Related Products
Sorry, an unexpected error has occurred
Response not successful: Received status code 500
__Related Articles__
- [Amino Acids Reference Chart](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/protein-biology/protein-structural-analysis/amino-acid-reference-chart)
- [Unnatural Amino Acids for Peptide Synthesis](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/chemistry-and-synthesis/peptide-synthesis/unnatural-amino-acids)
- [L-Glutamine in Cell Culture](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/cell-culture-and-cell-culture-analysis/mammalian-cell-culture/glutamine)
- [View More](https://www.sigmaaldrich.com/US/en/search/facet-search?focus=sitecontent&term=facet-search)
__Related Product Categories__
- [Amino Acids – Building Blocks of Life Science](https://www.sigmaaldrich.com/US/en/products/chemistry-and-biochemicals/biochemicals/amino-acids)
- [Amino Acid, Peptide & Protein Standards](https://www.sigmaaldrich.com/US/en/products/analytical-chemistry/reference-materials/amino-acid-peptide-and-protein-standards)
- [Amino Acids, Resins & Reagents for Peptide Synthesis](https://www.sigmaaldrich.com/US/en/products/chemistry-and-biochemicals/amino-acids-resins-and-reagents-for-peptide-synthesis)
- [Protection/Deprotection Reagents](https://www.sigmaaldrich.com/US/en/products/chemistry-and-biochemicals/chemical-synthesis/protection-deprotection-reagents)
- [pH Adjustment and Buffers](https://www.sigmaaldrich.com/US/en/products/pharma-and-biopharma-manufacturing/buffer-media-prep/buffers-and-ph-adjusters)
Top
__Sign In To Continue__
Sign In__Don't Have An Account?__Register
Support[Customer Support](https://www.sigmaaldrich.com/US/en/support/customer-support)[Contact Us](https://www.sigmaaldrich.com/US/en/collections/offices)[FAQ](https://maestro.my.site.com/knowledgeportal/s/)[Safety Data Sheets (SDS)](https://www.sigmaaldrich.com/US/en/documents-search?tab=sds)[Certificates (COA/COO)](https://www.sigmaaldrich.com/US/en/documents-search?tab=coa)[Quality & Regulatory](https://www.sigmaaldrich.com/US/en/life-science/quality-and-regulatory-management)[Calculators & Apps](https://www.sigmaaldrich.com/US/en/support/calculators-and-apps)[Webinars](https://www.sigmaaldrich.com/US/en/collections/webinars)
Orders[Order Lookup](https://www.sigmaaldrich.com/US/en/order-lookup)[Quick Order](https://www.sigmaaldrich.com/US/en/quick-order)[Custom Products](https://www.sigmaaldrich.com/US/en/services/custom-products)[eCommerce Solutions](https://www.sigmaaldrich.com/US/en/life-science/ecommerce/ecommerce-solutions)
Company[About Us](https://www.sigmaaldrich.com/US/en/life-science/about-us)[Responsibility](https://www.sigmaaldrich.com/US/en/life-science/ssbi)[Events](https://www.sigmaaldrich.com/US/en/collections/events)[Press Releases](https://www.sigmaaldrich.com/US/en/collections/press)[Programs](https://www.sigmaaldrich.com/US/en/life-science/partnership-programs)[Careers](https://careers.merckgroup.com/global/en)[Offices](https://www.sigmaaldrich.com/US/en/collections/offices)
Social Media
[](https://www.linkedin.com/company/merck-life-science)
[](https://x.com/Merck_lifesci)
[](https://www.facebook.com/Merck.lifescience)
[](https://www.instagram.com/mercklifescience/)
[](https://www.youtube.com/@MerckLifeScience/featured)
MilliporeSigma
__Research. Development. Production.__
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
[](https://www.sigmaaldrich.com/US/en)
[Sigma-Aldrich® Solutions](https://www.sigmaaldrich.com/US/en/life-science/sigma-aldrich)[BioReliance® Solutions](https://www.sigmaaldrich.com/US/en/life-science/bioreliance)[Millipore® Solutions](https://www.sigmaaldrich.com/US/en/life-science/millipore)[SAFC® Solutions](https://www.sigmaaldrich.com/US/en/life-science/safc)[Milli-Q® Solutions](https://www.sigmaaldrich.com/US/en/life-science/milliq)[Supelco® Solutions](https://www.sigmaaldrich.com/US/en/life-science/supelco)
© 2026 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved, including Text and Data Mining for AI training and similar technologies.
Reproduction of any materials from the site is strictly forbidden without permission.
[Site Use Terms](https://www.sigmaaldrich.com/US/en/life-science/legal/site-use-terms)|[Privacy Policy](https://www.sigmaaldrich.com/US/en/life-science/legal/privacy-statement)|[General Terms and Conditions of Sale](https://www.sigmaaldrich.com/US/en/life-science/legal/terms-and-conditions)|[Copyright Consent](https://www.sigmaaldrich.com/US/en/life-science/legal/copyright-consent) |[Site Map](https://www.sigmaaldrich.com/US/en/site-map)
An unknown error has occured.