[Skip to Content](https://www.sigmaaldrich.com#main-content)
[](https://www.sigmaaldrich.com/US/en)
Products
Cart0
USEN
Products
ProductsApplicationsServicesResourcesSupport
[Login / Register](https://www.sigmaaldrich.com/oidc-sign-in)
[Order Lookup](https://www.sigmaaldrich.com/US/en/order-lookup)
[Quick Order](https://www.sigmaaldrich.com/US/en/quick-order)
Cart0
[Home](https://www.sigmaaldrich.com/US/en)[Solid State Synthesis](https://www.sigmaaldrich.com/US/en/applications/materials-science-and-engineering/solid-state-synthesis)Nanostructured Materials via Ultrasonic Spray Pyrolysis
# Nanostructured Materials via Ultrasonic Spray Pyrolysis
__Jin Ho Bang1,2, Yuri T. Didenko3, Richard J. Helmich1,4, Kenneth S. Suslick1\*__
1School of Chemical Sciences, University of Illinois at Urbana-Champaign, 600 S. Mathews Av., Urbana, IL 61801 USA, 2Address: Department of Chemistry and Applied Chemistry, Hanyang University, 55 Hanyangdaehak-ro, Sangnok-gu, Ansan, Kyeonggi-do 426-791, Republic of Korea, 3UT Dots, Inc., 2716 W. Clark Rd, Suite E, Champaign, IL 61822 USA, 4Address: Dept. of Labor, Mine Safety and Health Administration, 6th Ave. and Kipling Pkwy, Bldg 25, Denver, CO 80225
[ksuslick@illinois.edu](mailto:ksuslick@illinois.edu)
## Introduction
Advances in nanostructured materials have often been led by the development of synthetic methods that provide control over size, morphology and structure. The preparation of materials in a scalable and continuous manner is critical when development moves beyond lab-scale quantities. The ease and scalability for preparing materials has been a driving force for the development of new methodologies for several decades.1 In this respect, the development of aerosol syntheses has been particularly successful.2 Spray pyrolysis has been widely used in industry for preparing fine powders, in addition to thin film deposition. This technique has been generally accepted because the apparatus is simple, continuous, and easily scaled up to mass production. In general, spray pyrolysis involves the thermal reactions of aerosols (i.e., solid or liquid droplets suspended in a gas) generated by a nebulizer (e.g. pneumatic, ultrasonic, or electrostatic) carried in a gas flow through a furnace.
Among the various nebulization techniques available (__Table 1__), the use of ultrasonic nebulizers has been favored because of their outstanding energy efficiency in aerosol generation, affordability, and the inherent low velocity of the initially formed aerosol. Ultrasonic spray pyrolysis (USP) has been shown to be particularly effective for the preparation of a wide array of materials, and it has the advantage of being highly scalable and continuous.3,4 Like sol gel process, USP technique is also a solution based method wherein high frequency ultrasound is passed through a liquid precursor solution, impinging on a liquid-gas interface, which forms an aerosol of micron-sized liquid droplets. The ultrasonically generated droplets of reaction precursors thus become individual chemical microreactors as they are carried by gas flow into a heated furnace where reactions occur. Evaporation of volatile solvent can be followed by chemical reactions either of the remaining solid or, in the presence of high boiling co-solvents or molten salts, in submicron-sized liquid droplets. Such reactions can occur in the interior of the droplet or on the droplet surface. A wide range of nanomaterials have been prepared by USP, including nanoporous metal oxides and sulfides, nanocomposites, high surface area carbons, semiconductor quantum dots, and conductive metal inks.
Atomizer Type
Average Initial
Droplet SizeDroplet
DistributionGas
VelocityDroplet
Delivery Rate
__Ultrasonic (submerged)__
1 to 10 μm
narrowlowmedium
__Ultrasonic (nozzle)__10 to 1000 μmmediumlowmedium to high
__Electrospray__<1 to 1000 μmnarrowlowlow to medium
__Rotary__10 to 1000 μmbroadlowhigh
__Pressure__10 to 1000 μmbroadhighhigh
Table 1 Characteristics of Various Nebulization Techniques.
## Ultrasonic Nebulization and Aerosol Formation
In ultrasonic spray pyrolysis, ultrasound does not induce reactions such as those commonly found in sonochemical reactions,4,5 but instead generates a low-velocity aerosol.6 The ultrasound used for these techniques is typically low intensity and high frequency, ~2 MHz. The droplet formation by ultrasound was first described by R. W. Wood and A. L. Loomis in 1927, and in 1962, Lang experimentally established the relationship between ultrasonic frequency and average droplet size:
where Dd is average droplet diameter, γ (N m-1) is the liquid’s surface tension, ρ (kg m-3) is the solution density, and *f*(MHz) is the ultrasonic frequency.[](https://www.sigmaaldrich.com/US/en/product/sial/208566)
Ultrasonic aerosol fountains are the result of capillary waves (i.e., waves travelling along the interface between two fluids) at the surface of liquids, and the aerosol generation is a result of momentum transfer. When the amplitude of the surface capillary waves is sufficiently high, the crests (peaks) of the waves can break off, resulting in liquid droplets. Since capillary wavelength is inversely proportional to wavelength, smaller droplets can be generated with higher frequencies.
## USP and CAFS Apparatus
A typical USP reactor is composed of a high frequency ultrasonic transducer (like that found in a household humidifier) submerged in water below the base of a nebulization vessel containing the precursor solution. A thin, flexible polymer film (e.g. polyethylene, polypropylene, or polytetrafluoroethylene) is used at the base of the reactor reservoir to separate the precursor solutions from the ultrasonic transducer. This configuration allows ultrasound to be effectively transmitted through the polymer membrane into the precursor solution allowing nebulization to occur, while sequestering the solution under an inert or reactive gas of one’s choosing. The nebulization vessel is fitted with a gas stream to carry the aerosol into a tubular furnace, and out to a collector (e.g., a bubbler, filter, electrostatic precipitator, etc.).
The USP process involves the sequence of (1) droplet generation, (2) evaporation of solvents in the heated zone, (3) diffusion of reactants, (4) reaction or precipitation, and (5) escape of any product volatiles. The nature of the final state is critically dependent on the phase of the material that remains after loss of the volatile solvent. USP has most commonly been used to produce solid particulates (i.e., spray drying) that may undergo solid state pyrolysis while in the tube furnace. More recently, several groups have used high boiling co-solvents, ionic liquids, or molten salts to keep the droplets of the reacting aerosol in a liquid state throughout the heated zone; such droplets then serve as isolated micron-sized chemical reactors. The use of liquid droplets has been referred to as salt-assisted7,8 USP or more generally, as chemical aerosol flow synthesis (CAFS).4,9-11
## Nanostructured Materials via Ultrasonic Spray Pyrolysis
### Metal Oxides
Historically, the formation of metal oxides by USP has been heavily studied.2,12 By nebulizing precursor solutions containing water soluble metal salts (e.g. chloride, nitrate, acetate, hydroxides) or metal coordination complexes (e.g. peroxo-, ammonium, lactate, oxo-), a myriad of mono- and multi-metallic oxide powders have been created by USP. With the addition of polymers, colloidal silica, or other composites forming materials, new and complex architectures can be easily added to many of the simple oxide particles previously reported. For instance, utilizing an aqueous solution containing a titanium complex and colloidal silica, Suh and Suslick have prepared various forms of nanostructured titania.13-14 This combination of precursors produces porous titania microspheres after selectively etching the silica template with HF. Interestingly, the addition of transition metal ions to the precursor solution caused an unusual phase separation to occur during USP, resulting in the outer shell becoming enriched in titania with the core of the particles consisting of silica. Etching the transition metal containing composites with HF for short periods of time yielded particles with a ball-in-ball morphology, and further etching completely removed the silica leaving a hollow, porous titania spheres behind.14 Cytotoxicity studies revealed the microspheres had little effect on the viability of various cell lines. Further studies using these materials for delivery of a potent drug for the treatment of Alzheimer’s disease (dehydroevodiamine) showed excellent selectivity for delivery to the cell cytosol rather than cell nuclei.15
Other examples of nanostructured oxide composites created by USP were pioneered by Okuyama and coworkers using polymer beads in the nebulized droplets as templates for creating macroporous oxide particles.16 One may expand upon this work by using inexpensive organic monomers (e.g., styrene), instead of expensive polymer beads, to generate interesting macroporous morphologies.13 One may also include Co2(CO)8 (Product No. [60811](https://www.sigmaaldrich.com/US/en/product/aldrich/60811)) in the precursor solution to form ferromagnetic microsphere composites with nanoparticles of Co encapsulated in silica; these show no loss of magnetic susceptibility due to the oxidation of Co even after 6 months.13 More recently, the ultrasonic spray synthesis of oxides has been aimed at creating materials with specific applications, such as sensors for the detection of toxic industrial gases and photocatalysts for solar energy conversion.17 For example, chemically responsive inks, which have important applications to chemical sensing, have been made using a CAFS method by the condensation reaction of tetramethoxysilane (Product No. [679259](https://www.sigmaaldrich.com/US/en/product/aldrich/679259), [341436](https://www.sigmaaldrich.com/US/en/product/aldrich/341436)) and methyltrimethoxysilane (Product No. [679232](https://www.sigmaaldrich.com/US/en/product/aldrich/679232), [246174](https://www.sigmaaldrich.com/US/en/product/aldrich/246174), [440175](https://www.sigmaaldrich.com/US/en/product/aldrich/440175)) around soluble dye molecules, producing an organically modified silica (ORMOSIL) pigment within the aerosol droplets.11
### Metal Sulfides and Other Chalcogenides
While ultrasonic spray has proven to be a versatile route to many different oxide materials, a variety of nanostructured sulfides and other chalcogenides can be prepared using both USP and CAFS methods. For example, Skrabalak and Suslick,18 pyrolyzed aerosol solutions of (NH4)2MoS4 (Product No. [323446](https://www.sigmaaldrich.com/US/en/product/aldrich/323446)) and prepared a highly effective MoS2 (Product No. [234842](https://www.sigmaaldrich.com/US/en/product/aldrich/234842), [69860](https://www.sigmaaldrich.com/US/en/product/aldrich/69860)) catalyst for hydrodesulfurization (HDS), the critical process for the removal of sulfur from petroleum. Porosity was induced in the MoS2 microspheres by adding colloidal silica to the precursor solution and subjecting the material to a post-synthesis etching step. MoS2 is a layered material with S-Mo-S---S-Mo-S layers; the catalytically active sites, however, are only the exposed Mo atoms at the edges of these layers. The nanoporosity induced by the silica template dramatically increases the number of active edge sites and the catalytic activity for hydrodesulfurization. When the porous MoS2 was salt impregnated with Co as a promoter, the activity increased well above that of RuS2, which is known to be the most intrinsically active metal sulfide for the HDS process.
CAFS has been used to prepare, on large scale, metal chalcogenide quantum dots (QDs).9,19 Didenko and Suslick developed a novel CAFS method using simple, low toxicity cadmium and chalcogenides precursors nebulized with a surfactant capping agent in a combination of toluene and high boiling point solvents (e.g., trioctylphosphine oxide). In this way, the individual droplets of solution act as reactors forming surfactant passivated CdS (Product No. [662550](https://www.sigmaaldrich.com/US/en/product/aldrich/662550)), CdSe (Product No. [662356](https://www.sigmaaldrich.com/US/en/product/aldrich/662356)), and CdTe (Product No. [716669](https://www.sigmaaldrich.com/US/en/product/aldrich/716669)) QDs. Compared to prior batch processing, which use large volumes of high boiling point solvents into which aggressive and toxic chemicals must be injected quickly and reproducibly, the CAFS method provides a useful flow reactor alternative. The micron-sized droplets of the CAFS precursors are sufficiently small that fast, uniform heating occurs and high quality QDs are obtained as the volatile toluene co-solvent rapidly evaporates. The QDs prepared by CAFS were highly fluorescence with a quantum yield of ~25%, and were tunable over a broad spectra region by simply changing the temperature of the furnace.
### Carbon Materials
Porous carbon materials are of interest for their potential use in electrochemical, catalytic, adsorbent, and gas-storage applications. Until recently, most porous carbons were prepared by carbonization of raw natural materials. Recent research efforts emphasize the use of templates and their subsequent removal to produce meso- and macroporous carbons with controlled, and in some cases, periodic pores. With this approach, a carbon precursor/inorganic template composite is first formed, followed by carbonization, then chemical leaching of the template material. Such methodology is tedious, requiring multiple synthetic steps, caustic chemical treatments, and long curing times; scale-up has also proven difficult and is not cost effective due to the destruction of (relatively) expensive templates.
CAFS provides an alternative approach for adding porosity, using molten salts within the droplets to form porous nanostructured carbon as reaction gases are released during the carbon formation. Skrabalak, Fortunato, and Suslick found that by nebulizing aqueous solutions of various organic salts (e.g. alkali metal chloroacetate and dichloroacetate) or even simple sugar mixed with alkali carbonate salts and pyrolyzing the aerosol droplets, they were able to create microspheres of carbon with diverse morphologies.20-21 The choice of precursors is based on compounds in which a single carbon remains after the likely leaving groups (e.g., CO2, HCl, H2O) have been pyrolyzed away. These carbon materials have proved useful as catalyst supports in direct methanol fuel cells (DMFCs).22 By loading the porous carbons created by USP with catalysts consisting of PtRu (Product No. [738573](https://www.sigmaaldrich.com/US/en/product/aldrich/738573)) and Pt (Product No. [204013](https://www.sigmaaldrich.com/US/en/product/aldrich/204013), [327441](https://www.sigmaaldrich.com/US/en/product/aldrich/327441), [327476](https://www.sigmaaldrich.com/US/en/product/aldrich/327476)) alone for the anode and cathode, respectively, it was shown that the USP carbon microspheres could improve the electrical output of DMFCs when combined with traditional support materials in the anode and cathode.
### Metals
USP can also be used to prepare metal nanoparticles and nanostructures for many precious and nonprecious metals, almost always by the use of metal oxides that decompose at temperatures below 1000 °C or by high temperature reduction with hydrogen in the flow gas.23 Silver nanoparticles (of interest as printable conductive inks) (Product No. [675318](https://www.sigmaaldrich.com/US/en/product/aldrich/675318)) have been made by Hong and coworkers using a liquid phase microreactor for the thermal reduction of silver pentafluoropropionate ([326216](https://www.sigmaaldrich.com/US/en/product/aldrich/326216)) in isoamyl ether (Product No. [260649](https://www.sigmaaldrich.com/US/en/product/aldrich/260649)) in the presence of trioctylamine24 (Product No. [T81000](https://www.sigmaaldrich.com/US/en/product/aldrich/t81000)) and a similar process was then scaled up using USP by Didenko and Ni.19 Forming highly reactive metal nanostructures (i.e., metals whose standard reduction potential is less than hydrogen) has remained a serious challenge, however, one that Helmich and Suslick recently addressed in a CAFS synthesis of metallic aluminum (Product No. [518573](https://www.sigmaaldrich.com/US/en/product/aldrich/518573), [653608](https://www.sigmaaldrich.com/US/en/product/aldrich/653608)).25 Hollow, metallic aluminum particles were prepared by reacting trimethylamine aluminum hydride solution droplets with TiCl4 (Product No. [697079](https://www.sigmaaldrich.com/US/en/product/aldrich/697079), [208566](https://www.sigmaaldrich.com/US/en/product/sigald/208566), 254312) vapor. Depending on the solution concentration and reaction temperature the morphology of the hollow microspheres could be changed from macroporous to nonporous. Ultrafine and nanoparticles of metallic aluminum have historically found use in propellants and pyrotechnics26 and more recently as nanothermites.27 The hollow morphology of the aluminum particles prepared by CAFS could easily be developed into similar applications.
### Scale-Up and Industrial Applications
Ultrasonic spray pyrolysis on the laboratory scale at high frequencies (~2 MHz) as described earlier is useful for rapid hypothesis testing and can easily prepare new and exotic materials on a small scale. The ultrasound intensity on the surface of the high frequency transducer, however, is not sufficient to atomize viscous, dense, or high boiling solvents and slurries. In order to increase product throughput and spray larger amounts of liquid precursors, industrial scale setups for generation of small droplets of liquids using high intensity ultrasound are commercially available. Such ultrasonic nozzles work at frequencies from 20 to 120 kHz to create different droplet sizes (down to ~10 μm) and can spray up to ~100 mL/min of liquids with high viscosities, low vapor pressures and even slurries.19
The yield and the quality of final products depends on the gas and liquid flow rates through the reactor, temperature inside the reactor, ultrasound power, solvents used and chemical composition of the mixture of precursors. By varying parameters of the experiments, it is possible to get high yields. A variety of products can be obtained in this setup, including cadmium chalcogenide quantum dots (both core and core/shell), metal nanoparticles (silver, copper, nickel, copper/nickel and copper/silver alloys), and metal oxides. Many nanomaterials have been synthesized in a continuous-flow process using this apparatus.
The commercial market for quantum dots is currently dominated by biomedical applications and was worth of $67 million in 2010 and is projected to grow to $670 million by 2015.28 Biomedical applications include bioimaging, flow cytometry, fluorescent labeling for cancer detection and intracellular imaging. The optoelectronics market for quantum dots is small at present, but likely to grow substantially with development of quantum dots for large scale industrial applications including solar energy harvesting, optical filters, displays, LEDs, etc.
The USP production of 10 nm silver nanoparticles (Product No. [730785](https://www.sigmaaldrich.com/US/en/product/aldrich/730785)), and 4 nm gold nanoparticles have commercial applications as the base for high quality, inexpensive, low temperature sintering nanometallic inks, which opens the door for large-scale production of printable electronics as well as antimicrobial uses. The largest application of nanometals industrially is the nanosilver market, which is expected to grow from $290 million in 2011 to ~$1.2 billion in 2016.29-30 Nanosilver is used as an antibacterial material in filters for air and water purification, in textiles, and for wound treatment.30-31 Other large scale applications of nanosilver include conductive nanoinks for printed circuits, front-side metallization of solar cells, thin film electronics, sensors, and catalysts. In addition, the market for other nanometals, such as gold, platinum and palladium may grow at an even higher rate than silver, specifically for biomedical applications.
As a final example, there are widespread and increasingly large markets for nanostructured metal oxides, from biomedical to photovoltaics and photocatalysts to lithium batteries.30, 32-33 Nanostructured silica, titania, iron oxides, zinc oxides, and a variety of binary/ternary metal oxides can all be produced via USP,12-16 and they have a variety of therapeutic applications and some are finding new applications in drug delivery forumulations.33-35 Design of biodegradable and biocompatible nanocarriers based on these materials is under active development.31, 33-35
## Concluding Remarks and Future Outlook
Ultrasonic spray methods for preparing nanostructured materials have proven to be powerful tools and the revival of interest in USP over the past decade has been substantial. We have highlighted the diversity of applications and nanostructured materials accessible using ultrasonic spray methods. We expect that future work in this field will focus increasingly on the preparation of novel functional materials developed with specific purposes in mind (e.g., sorbents, catalyst and catalyst supports, sensors, and energetic materials) where the relationship between morphology and function can be readily pursued by exploring new precursors and aerosol synthetic routes.
## Acknowledgments
This work was supported in part by the NSF DMR 09-06904, OII-0539385, IIP-0637721, and IIP-0823029. Materials characterizations were carried out in the Center for Microanalysis of Materials at the University of Illinois at Urbana-Champaign, which is partially supported by the U.S. DOE under grants DE-FG02-07ER46453 and DE-FG02-07ER46471.
## Materials
Sorry, an unexpected error has occurred
Response not successful: Received status code 500
### References
1\.
Hummel RE. 2004. Understanding Materials Science. [https://doi.org/10.1007/b137957](https://doi.org/10.1007/b137957)
2\.
Toivo Tarmo K, Toivo T. K, Hampden-Smith MJ. 1999 . Aerosol Processing of Materials , . NY Wiley: University of Michigan.
3\.
Nandiyanto ABD, Okuyama K. 2011. Progress in developing spray-drying methods for the production of controlled morphology particles: From the nanometer to submicrometer size ranges. Advanced Powder Technology. 22(1):1-19. [https://doi.org/10.1016/j.apt.2010.09.011](https://doi.org/10.1016/j.apt.2010.09.011)
4\.
Bang JH, Suslick KS. Applications of Ultrasound to the Synthesis of Nanostructured Materials. Adv. Mater.. 22(10):1039-1059. [https://doi.org/10.1002/adma.200904093](https://doi.org/10.1002/adma.200904093)
5\.
Suslick KS, Price GJ. 1999. APPLICATIONS OF ULTRASOUND TO MATERIALS CHEMISTRY. Annu. Rev. Mater. Sci.. 29(1):295-326. [https://doi.org/10.1146/annurev.matsci.29.1.295](https://doi.org/10.1146/annurev.matsci.29.1.295)
6\.
Ashgriz N. 2011. Handbook of Atomization and Sprays. [https://doi.org/10.1007/978-1-4419-7264-4](https://doi.org/10.1007/978-1-4419-7264-4)
7\.
Xia B, Lenggoro I, Okuyama K. 2001. Novel route to nanoparticle synthesis by salt‐assisted aerosol decomposition Advanced Materials . 13(20):1579-1582.
8\.
Okuyama K, Wuled Lenggoro I. 2003. Preparation of nanoparticles via spray route. Chemical Engineering Science. 58(3-6):537-547. [https://doi.org/10.1016/s0009-2509(02)00578-x](https://doi.org/10.1016/s0009-2509%2802%2900578-x)
9\.
Didenko YT, Suslick KS. 2005. Chemical Aerosol Flow Synthesis of Semiconductor Nanoparticles. J. Am. Chem. Soc.. 127(35):12196-12197. [https://doi.org/10.1021/ja054124t](https://doi.org/10.1021/ja054124t)
10\.
Bang JH, Suh WH, Suslick KS. 2008. Quantum Dots from Chemical Aerosol Flow Synthesis: Preparation, Characterization, and Cellular Imaging. Chem. Mater.. 20(12):4033-4038. [https://doi.org/10.1021/cm800453t](https://doi.org/10.1021/cm800453t)
11\.
Bang JH, Lim SH, Park E, Suslick KS. 2008. Chemically Responsive Nanoporous Pigments: Colorimetric Sensor Arrays and the Identification of Aliphatic Amines. Langmuir. 24(22):13168-13172. [https://doi.org/10.1021/la802029m](https://doi.org/10.1021/la802029m)
12\.
Messing GL, Zhang S, Jayanthi GV. 1993. Ceramic Powder Synthesis by Spray Pyrolysis. J American Ceramic Society. 76(11):2707-2726. [https://doi.org/10.1111/j.1151-2916.1993.tb04007.x](https://doi.org/10.1111/j.1151-2916.1993.tb04007.x)
13\.
Suh WH, Suslick KS. 2005. Magnetic and Porous Nanospheres from Ultrasonic Spray Pyrolysis. J. Am. Chem. Soc.. 127(34):12007-12010. [https://doi.org/10.1021/ja050693p](https://doi.org/10.1021/ja050693p)
14\.
Suh W, Jang A, Suh Y, Suslick K. 2006. Porous, Hollow, and Ball-in-Ball Metal Oxide Microspheres: Preparation, Endocytosis, and Cytotoxicity. Adv. Mater.. 18(14):1832-1837. [https://doi.org/10.1002/adma.200600222](https://doi.org/10.1002/adma.200600222)
15\.
Suh W, Suslick K, Suh Y. 2005. Therapeutic Agents for Alzheimers Disease. CMCCNSA. 5(4):259-269. [https://doi.org/10.2174/156801505774913035](https://doi.org/10.2174/156801505774913035)
16\.
Iskandar F, Mikrajuddin, Okuyama K. 2002. Controllability of Pore Size and Porosity on Self-Organized Porous Silica Particles. Nano Lett.. 2(4):389-392. [https://doi.org/10.1021/nl015662g](https://doi.org/10.1021/nl015662g)
17\.
Bang JH, Helmich RJ, Suslick KS. 2008. Nanostructured ZnS:Ni2+Photocatalysts Prepared by Ultrasonic Spray Pyrolysis. Adv. Mater.. 20(13):2599-2603. [https://doi.org/10.1002/adma.200703188](https://doi.org/10.1002/adma.200703188)
18\.
Skrabalak SE, Suslick KS. 2005. Porous MoS2Synthesized by Ultrasonic Spray Pyrolysis. J. Am. Chem. Soc.. 127(28):9990-9991. [https://doi.org/10.1021/ja051654g](https://doi.org/10.1021/ja051654g)
19\.
Didenko Y, Ni Y. 2010. Controlled Synthesis of Nanoparticles Using Continuous Liquid-Flow Aerosol Method US Patent 7,811,543 B2 issued 10/12/2010.
20\.
Skrabalak SE, Suslick KS. 2006. Porous Carbon Powders Prepared by Ultrasonic Spray Pyrolysis. J. Am. Chem. Soc.. 128(39):12642-12643. [https://doi.org/10.1021/ja064899h](https://doi.org/10.1021/ja064899h)
21\.
Fortunato ME, Rostam-Abadi M, Suslick KS. 2010. Nanostructured Carbons Prepared by Ultrasonic Spray Pyrolysis. Chem. Mater.. 22(5):1610-1612. [https://doi.org/10.1021/cm100075j](https://doi.org/10.1021/cm100075j)
22\.
Bang JH, Han K, Skrabalak SE, Kim H, Suslick KS. 2007. Porous Carbon Supports Prepared by Ultrasonic Spray Pyrolysis for Direct Methanol Fuel Cell Electrodes. J. Phys. Chem. C. 111(29):10959-10964. [https://doi.org/10.1021/jp071624v](https://doi.org/10.1021/jp071624v)
23\.
Gürmen S, Stopi? S, Friedrich B. 2006. Synthesis of nanosized spherical cobalt powder by ultrasonic spray pyrolysis. Materials Research Bulletin. 41(10):1882-1890. [https://doi.org/10.1016/j.materresbull.2006.03.006](https://doi.org/10.1016/j.materresbull.2006.03.006)
24\.
Lin XZ, Terepka AD, Yang H. 2004. Synthesis of Silver Nanoparticles in a Continuous Flow Tubular Microreactor. Nano Lett.. 4(11):2227-2232. [https://doi.org/10.1021/nl0485859](https://doi.org/10.1021/nl0485859)
25\.
Helmich RJ, Suslick KS. 2010. Chemical Aerosol Flow Synthesis of Hollow Metallic Aluminum Particles. Chem. Mater.. 22(17):4835-4837. [https://doi.org/10.1021/cm101342r](https://doi.org/10.1021/cm101342r)
26\.
Russell MS. 2009 . The Chemistry of Fireworks Cambridge Royal Society of Chemistry.
27\.
Puszynski JA. 2009. Processing and characterization of aluminum-based nanothermites. J Therm Anal Calorim. 96(3):677-685. [https://doi.org/10.1007/s10973-009-0037-0](https://doi.org/10.1007/s10973-009-0037-0)
28\.
Oliver J. 2011 . Quantum Dots: Global Market Growth and Future Commercial Prospects . Wellesley: MA BCC Research.
29\.
2012\. Nanometals in Electronics and Energy Markets and Beyond NanoMarkets.net . \[Internet]. VA : Glen Allen.\[cited 20 Dec 2011]. Available from: [https://www.prnewswire.com/news-releases/nanometals-in-electronics-and-energy-markets--2012-and-beyond-136580283.html](https://www.prnewswire.com/news-releases/nanometals-in-electronics-and-energy-markets--2012-and-beyond-136580283.html)
30\.
Evers P. 2012 . Nanotechnology in Medical Applications: The Global Market. Wellesley: MA BCC Research.
31\.
Li Y, Chen C. 2011. Fate and Toxicity of Metallic and Metal-Containing Nanoparticles for Biomedical Applications. Small. 7(21):2965-2980. [https://doi.org/10.1002/smll.201101059](https://doi.org/10.1002/smll.201101059)
32\.
Ji L, Lin Z, Alcoutlabi M, Zhang X. 2011. Recent developments in nanostructured anode materials for rechargeable lithium-ion batteries. Energy Environ. Sci.. 4(8):2682. [https://doi.org/10.1039/c0ee00699h](https://doi.org/10.1039/c0ee00699h)
33\.
Kumar C. 2009 . Nanostructured Oxides Weinheim Wiley-VCH.
34\.
Chandra S, Barick K, Bahadur D. 2011. Oxide and hybrid nanostructures for therapeutic applications. Advanced Drug Delivery Reviews. 63(14-15):1267-1281. [https://doi.org/10.1016/j.addr.2011.06.003](https://doi.org/10.1016/j.addr.2011.06.003)
35\.
Suh WH, Suslick KS, Stucky GD, Suh Y. 2009. Nanotechnology, nanotoxicology, and neuroscience. Progress in Neurobiology. 87(3):133-170. [https://doi.org/10.1016/j.pneurobio.2008.09.009](https://doi.org/10.1016/j.pneurobio.2008.09.009)
__Related Articles__
- [Role of High Purity Metal Salts in Chemical Synthesis](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/chemistry-and-synthesis/ch-functionalization/role-of-high-purity-metal-salts-in-chemical-synthesis)
- [Complex Hydrides: New Solid-state Conductors](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/materials-science-and-engineering/batteries-supercapacitors-and-fuel-cells/complex-hydrides)
- [Mesoporous Oxides and Their Applications to Hydrogen Storage](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/materials-science-and-engineering/batteries-supercapacitors-and-fuel-cells/mesoporous-oxides)
- [Nano Olivine-based Cathode Materials for Li-ion Batteries](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/materials-science-and-engineering/batteries-supercapacitors-and-fuel-cells/nanostructured-olivine-based-cathode-materials)
- [Intermetallic Hydrides with High Dissociation Pressure](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/materials-science-and-engineering/batteries-supercapacitors-and-fuel-cells/nanostructured-olivine-based-cathode-materials/intermetallic-hydrides-with-high-dissociation-pressure)
- [Molecular Monolayers on Silicon Surfaces](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/materials-science-and-engineering/biosensors-and-imaging/molecular-monolayers)
- [Molybdenum Disulfide: Hydrogen Evolution](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/materials-science-and-engineering/chemical-vapor-deposition/molybdenum-disulfide)
- [Innovative Materials for Glass Ionomer Cements (GICs): Dental Applications](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/materials-science-and-engineering/contact-lens-and-dental-manufacturing/innovative-ionomers-for-dental-applications)
- [View More](https://www.sigmaaldrich.com/US/en/search/facet-search?focus=sitecontent&term=facet-search)
__Related Product Categories__
- [Hydrogenation Catalysts](https://www.sigmaaldrich.com/US/en/products/chemistry-and-biochemicals/catalysts/hydrogenation-catalysts)
- [Organosilicon Reagents](https://www.sigmaaldrich.com/US/en/products/chemistry-and-biochemicals/chemical-synthesis/organosilicon-reagents)
- [Inorganic Catalysts](https://www.sigmaaldrich.com/US/en/products/chemistry-and-biochemicals/catalysts/inorganic-catalysts)
- [Organic Building Blocks](https://www.sigmaaldrich.com/US/en/products/chemistry-and-biochemicals/building-blocks/organic-building-blocks)
- [Caustic alkalis and bases](https://www.sigmaaldrich.com/US/en/products/chemistry-and-biochemicals/lab-chemicals/caustic-alkalis-and-bases)
- [Polymerization Tools](https://www.sigmaaldrich.com/US/en/products/materials-science/biomedical-materials/polymerization-tools)
- [Salts](https://www.sigmaaldrich.com/US/en/products/chemistry-and-biochemicals/lab-chemicals/salts)
Top
__Sign In To Continue__
Sign In__Don't Have An Account?__Register
Support[Customer Support](https://www.sigmaaldrich.com/US/en/support/customer-support)[Contact Us](https://www.sigmaaldrich.com/US/en/collections/offices)[FAQ](https://maestro.my.site.com/knowledgeportal/s/)[Safety Data Sheets (SDS)](https://www.sigmaaldrich.com/US/en/documents-search?tab=sds)[Certificates (COA/COO)](https://www.sigmaaldrich.com/US/en/documents-search?tab=coa)[Quality & Regulatory](https://www.sigmaaldrich.com/US/en/life-science/quality-and-regulatory-management)[Calculators & Apps](https://www.sigmaaldrich.com/US/en/support/calculators-and-apps)[Webinars](https://www.sigmaaldrich.com/US/en/collections/webinars)
Orders[Order Lookup](https://www.sigmaaldrich.com/US/en/order-lookup)[Quick Order](https://www.sigmaaldrich.com/US/en/quick-order)[Custom Products](https://www.sigmaaldrich.com/US/en/services/custom-products)[eCommerce Solutions](https://www.sigmaaldrich.com/US/en/life-science/ecommerce/ecommerce-solutions)
Company[About Us](https://www.sigmaaldrich.com/US/en/life-science/about-us)[Responsibility](https://www.sigmaaldrich.com/US/en/life-science/ssbi)[Events](https://www.sigmaaldrich.com/US/en/collections/events)[Press Releases](https://www.sigmaaldrich.com/US/en/collections/press)[Programs](https://www.sigmaaldrich.com/US/en/life-science/partnership-programs)[Careers](https://careers.merckgroup.com/global/en)[Offices](https://www.sigmaaldrich.com/US/en/collections/offices)
Social Media
[](https://www.linkedin.com/company/merck-life-science)
[](https://x.com/Merck_lifesci)
[](https://www.facebook.com/Merck.lifescience)
[](https://www.instagram.com/mercklifescience/)
[](https://www.youtube.com/@MerckLifeScience/featured)
MilliporeSigma
__Research. Development. Production.__
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
[](https://www.sigmaaldrich.com/US/en)
[Sigma-Aldrich® Solutions](https://www.sigmaaldrich.com/US/en/life-science/sigma-aldrich)[BioReliance® Solutions](https://www.sigmaaldrich.com/US/en/life-science/bioreliance)[Millipore® Solutions](https://www.sigmaaldrich.com/US/en/life-science/millipore)[SAFC® Solutions](https://www.sigmaaldrich.com/US/en/life-science/safc)[Milli-Q® Solutions](https://www.sigmaaldrich.com/US/en/life-science/milliq)[Supelco® Solutions](https://www.sigmaaldrich.com/US/en/life-science/supelco)
© 2026 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved, including Text and Data Mining for AI training and similar technologies.
Reproduction of any materials from the site is strictly forbidden without permission.
[Site Use Terms](https://www.sigmaaldrich.com/US/en/life-science/legal/site-use-terms)|[Privacy Policy](https://www.sigmaaldrich.com/US/en/life-science/legal/privacy-statement)|[General Terms and Conditions of Sale](https://www.sigmaaldrich.com/US/en/life-science/legal/terms-and-conditions)|[Copyright Consent](https://www.sigmaaldrich.com/US/en/life-science/legal/copyright-consent) |[Site Map](https://www.sigmaaldrich.com/US/en/site-map)