Bar to PSI Conversion Chart
Convert Bar to PSI
Use this bar to psi conversion table and chart for your own use in the lab or field. Easily convert bars to psi quickly and simply. Designed for easy conversions of bar to psi. Convert psi to bars using the table in reverse. For research, laboratory work or field use. 
Formula for bar to psi conversion: 1 bar = 14.5038 psi or (bar x 14.5038 = psi) or 0.0689475729 bars in 1 psi.
What is psi? psi stands for pounds per square inch or pounds of force per square inch.
Bar psi
1 14.50377377
1.1 15.95415115
1.2 17.40452853
1.3 18.8549059
1.4 20.30528328
1.5 21.75566066
1.6 23.20603804
1.7 24.65641541
1.8 26.10679279
1.9 27.55717017
2 29.00754755
2.25 32.63349099
2.5 36.25943443
2.75 39.88537788
3 43.51132132
3.25 47.13726476
3.5 50.76320821
3.75 54.38915165
4 58.01509509
4.25 61.64103854
4.5 65.26698198
4.75 68.89292542
5 72.51886887
6 87.02264264
7 101.5264164
8 116.0301902
9 130.533964
10 145.0377377
11 159.5415115
12 174.0452853
13 188.549059
14 203.0528328
15 217.5566066
20 290.0754755
25 362.5943443
30 435.1132132
35 507.6320821
40 580.1509509
45 652.6698198
50 725.1886887
55 797.7075575
60 870.2264264
65 942.7452952
70 1015.264164
75 1087.783033
80 1160.301902
85 1232.820771
90 1305.33964
95 1377.858508
100 1450.377377
105 1522.896246
110 1595.415115
115 1667.933984
120 1740.452853
125 1812.971722
150 2175.566066
175 2538.16041
200 2900.754755
225 3263.349099
250 3625.943443
275 3988.537788
300 4351.132132
325 4713.726476
350 5076.320821
375 5438.915165
400 5801.509509