[Skip to Content](https://www.sigmaaldrich.com#main-content)
[](https://www.sigmaaldrich.com/US/en)
Products
Cart0
USEN
Products
ProductsApplicationsServicesResourcesSupport
[Login / Register](https://www.sigmaaldrich.com/oidc-sign-in)
[Order Lookup](https://www.sigmaaldrich.com/US/en/order-lookup)
[Quick Order](https://www.sigmaaldrich.com/US/en/quick-order)
Cart0
[Home](https://www.sigmaaldrich.com/US/en)[Drinking Water Testing](https://www.sigmaaldrich.com/US/en/applications/environmental-testing-and-industrial-hygiene/drinking-water-testing)Safe Drinking Water Act & Clean Water Act – What is it?
# Safe Drinking Water Act & Clean Water Act – What is it?
## Safe Drinking Water Act
The Safe Drinking Water Act reflects the primary drinking water regulation. Drinking water here means finished drinking water from the plant and the distribution lines. But the Safe Drinking Water Act covers also the ground water sources for raw water and the surface water sources for Raw Drinking Water.
## Clean Water Act
The Clean Water Act reflects the wastewater analyses. But it is not only the wastewater. The Clean Water Act covers also the Storm Water, the Surface Fresh Water like Rivers, Lakes and Streams. Also, the Marine Water with the control of the bays, the Estuaries and the Beaches are covered by the Clean Water Act too.
## Documents for Safe Drinking Water and Clean Water Act
We continue to expand the portfolio of products to support the testing needs of environmental labs in the US for both Clean Water Act and Safe Drinking Water Act testing. A major component of this expansion is the validation and reporting of the spectrophotometric test kits used for analytes of interest. This page contains all of the documentation and reporting needed by any lab to satisfy auditors of adequate quality control and product suitability.
In some cases, products were issued approval letters by the USEPA in the past. Due to policy changes at the USEPA, approval letters are no longer provided and instead vendors and labs have to provide adequate documentation of equivalency. For those products where equivalency documentation is required, we have provided certificates of compliance, single laboratory validation and a helpful checklist for labs to comply with the guidelines established in the 2007 EPA Memo from Richard Reding.
We will continue to add and expand the product reports on this page as they become available. Please contact us if there is a specific product you would like more information on.
## Approvals / Equivalency Documents
Available for the following parameters:
- [Ammonia (Ammonium)](https://www.sigmaaldrich.com#ammonia)
- [Chlorine](https://www.sigmaaldrich.com#chlorine)
- [Chromate (Chromium VI)](https://www.sigmaaldrich.com#chromate)
- [COD](https://www.sigmaaldrich.com#cod)[](https://www.sigmaaldrich.com#cod)
- [](https://www.sigmaaldrich.com#cod)[Cyanide (not available in North America)](https://www.sigmaaldrich.com#cyanide)
- [Nitrate](https://www.sigmaaldrich.com#nitrate)
- [Nitrite](https://www.sigmaaldrich.com#nitrite)
- [Phosphate](https://www.sigmaaldrich.com#phosphate)
- [Sulfate](https://www.sigmaaldrich.com#sulfate)
We are working to provide you more.
USEPA Documentation for Spectroquant® Test Kits
### Ammonia (Ammonium)
USEPA Equivalency for
__Product No.____Spectroquant® Test Kits____Measuring Ranges____Clean Water Act____Safe Drinking Water Act__
[1.14558](https://www.sigmaaldrich.com/US/en/product/mm/114558)Ammonium Cell Test0.20 – 8.00 mg/L NH4-N
0.26 – 10.30 mg/L NH4+
0.20 – 8.00 mg/L NH3-N
0.26 – 9.73 mg/L NH3YesYes*
[1.14544](https://www.sigmaaldrich.com/US/en/product/mm/114544)Ammonium Cell Test0.5 – 16.0 mg/L NH4-N
0.6 – 20.6 mg/L NH4+
0.5 – 16.0 mg/L NH3-N
0.6 – 19.5 mg/L NH3 YesYes*
[1.14559](https://www.sigmaaldrich.com/US/en/product/mm/114559)Ammonium Cell Test4.0 – 80.0 mg/L NH4-N
5.2 – 103.0 mg/L NH4+
4.0 – 80.0 mg/L NH3-N
5.2 – 97.3 mg/L NH3 YesYes*
[1.14752](https://www.sigmaaldrich.com/US/en/product/mm/114752)Ammonium Test0.010 – 3.00 mg/L NH4-N
0.013 – 3.86 mg/L NH4+
0.010 – 3.00 mg/L NH3-N
0.013 – 3.65 mg/L NH3 YesYes*
[1.00683](https://www.sigmaaldrich.com/US/en/product/mm/100683)Ammonium Test2.0 – 150 mg/L NH4-N
2.6 – 193 mg/L NH4+
2.0 – 150 mg/L NH3-N
2.6 – 182 mg/L NH3YesYes*
\* Safe Drinking Water Act uses methods in 40 CFR part 136 Clean Water Act. The Merck KGaA, Darmstadt, Germany, Equivalency documents covers all pack sizes, cells and citation units mentioned in the insert sheet of the test kit.
### Chlorine
USEPA Equivalency for
__Product No.____Spectroquant® Test Kits____Measuring Ranges____Clean Water Act____Safe Drinking Water Act__
[1.00595](https://www.sigmaaldrich.com/US/en/product/mm/100595)Chlorine Cell Test (free chlorine)0.03 - 6.00 mg/L CI2 Yes
[1.00598](https://www.sigmaaldrich.com/US/en/product/mm/100598)Chlorine Test (free chlorine)0.010 - 6.00 mg/L Cl2 Yes
[1.00602](https://www.sigmaaldrich.com/US/en/product/mm/100602) Chlorine Test (total chlorine)0.010 - 6.00 mg/L Cl2YesYes
[1.00597](https://www.sigmaaldrich.com/US/en/product/mm/100597)Chlorine Cell Test (free and total chlorine)0.03 - 6.00 mg/L Cl2YesYes
[1.00599](https://www.sigmaaldrich.com/US/en/product/mm/100599)Chlorine Test for (free and total chlorine)0.010 - 6.00 mg/L Cl2YesYes
### Chromate (Chromium VI)
USEPA Equivalency for
__Product No.____Spectroquant® Test Kits____Measuring Ranges____Clean Water Act____Safe Drinking Water Act__
[1.14552](https://www.sigmaaldrich.com/US/en/product/mm/114552)Chromate Cell Test (Chromium VI)0.05 - 2.0 mg/L CrYes
### COD
USEPA Equivalency for
__Product No.____Spectroquant® Test Kits____Measuring Ranges____Clean Water Act____Safe Drinking Water Act__
[1.14560](https://www.sigmaaldrich.com/US/en/product/mm/114560)COD Cell Test 4.0 - 40.0 mg/L CODYes
[1.01796](https://www.sigmaaldrich.com/US/en/product/mm/101796)COD Cell Test 5.0 – 80.0 mg/L CODYes
[1.14540](https://www.sigmaaldrich.com/US/en/product/mm/114540)COD Cell Test 10 - 150 mg/L CODYes
[1.14895](https://www.sigmaaldrich.com/US/en/product/mm/114895)COD Cell Test 15 - 300 mg/L CODYes
[1.14541](https://www.sigmaaldrich.com/US/en/product/mm/114541)COD Cell Test 25 - 1,500 mg/L CODYes
[1.14691](https://www.sigmaaldrich.com/US/en/product/mm/114691)COD Cell Test 300 - 3,500 mg/L CODYes
[1.14555](https://www.sigmaaldrich.com/US/en/product/mm/114555)COD Cell Test 500 - 10,000 mg/L CODYes
[1.01797](https://www.sigmaaldrich.com/US/en/product/mm/101797)COD Cell Test 5,000 – 90,000 mg/L CODYes
### Cyanide
USEPA Equivalency for
__Product No.____Spectroquant® Test Kits____Measuring Ranges____Clean Water Act____Safe Drinking Water Act__
[1.14561](https://www.sigmaaldrich.com/US/en/product/mm/114561)Cyanide Cell Test (free cyanide)0.010 - 0.500 mg/L CNYes
### Nitrate
USEPA Equivalency for
__Product No.____Spectroquant® Test Kits____Measuring Ranges____Clean Water Act____Safe Drinking Water Act__
[1.14542](https://www.sigmaaldrich.com/US/en/product/mm/114542)Nitrate Cell Test 0.5-18.0 mg/L NO3-NYes
2.2 – 79.7 mg/L NO3
[1.14563](https://www.sigmaaldrich.com/US/en/product/mm/114563)Nitrate Cell Test 0.5 - 25.0 mg/L NO3-NYes
2.2 – 110.7 mg/L NO3
[1.14764](https://www.sigmaaldrich.com/US/en/product/mm/114764)Nitrate Cell Test 1.0 - 50.0 mg/L NO3-NYes
4 – 221 mg/L NO3
[1.00614](https://www.sigmaaldrich.com/US/en/product/mm/100614)Nitrate Cell Test 23-225 mg/L NO3-NYes
102 – 996 mg/L NO3
[1.09713](https://www.sigmaaldrich.com/US/en/product/mm/109713)Nitrate Test 0.10-25.0 mg/L NO3-NYes
0.4 – 110.7 mg/L NO3
[1.14773](https://www.sigmaaldrich.com/US/en/product/mm/114773)Nitrate Test 0.2-20.0 mg/L NO3-NYes
0.89 – 88.5 mg/L NO3
[1.01842](https://www.sigmaaldrich.com/US/en/product/mm/101842)Nitrate Test 0.3-30.0 mg/L NO3-NYes
1.3 – 132.8 mg/L NO3
[1.14556](https://www.sigmaaldrich.com/US/en/product/mm/114556)Nitrate Cell Test in seawater0.10-3.00 mg/L NO3-NYes
0.4 – 13.3 mg/L NO3
[1.14942](https://www.sigmaaldrich.com/US/en/product/mm/114942)Nitrate Test in seawater 0.2-17.0 mg/L NO3-NYes
0.9 – 75.3 mg/L NO3
### Nitrite
__Product No.____Spectroquant® Test Kits____Measuring Ranges____USEPA Equivalency for__
__Clean Water Act____Safe Drinking Water Act__
[1.14547](https://www.sigmaaldrich.com/US/en/product/mm/114547)Nitrite Cell Test0.010 – 0.700 mg/L NO2-N
0.03 – 2.30 mg/L NO2Yes
[1.00609](https://www.sigmaaldrich.com/US/en/product/mm/100609)Nitrite Cell Test1.0 – 90.0 mg/L NO2-N
3.0 – 295.2 mg/L NO2Yes
[1.14776](https://www.sigmaaldrich.com/US/en/product/mm/114776)Nitrite Test0.002 – 1.00 mg/L NO2-N
0.007 – 3.28 mg/L NO2
### Phosphate
USEPA Equivalency for
__Product No.____Spectroquant® Test Kits____Measuring Ranges____Clean Water Act____Safe Drinking Water Act__
[1.14543](https://www.sigmaaldrich.com/US/en/product/mm/114543)Phosphate Cell Test0.05 – 5.00 mg/L PO4-PYesYes
(orthophosphate ad Total phosphorus)0.2 – 15.3 mg/L PO4
[1.14729](https://www.sigmaaldrich.com/US/en/product/mm/114729)Phosphate Cell Test0.5 – 25.0 mg/L PO4-PYesYes
(orthophosphate ad Total phosphorus)1.5 – 76.7 mg/L PO4
### Sulfate
USEPA Equivalency for
__Product No.____Spectroquant® Test Kits____Measuring Ranges____Clean Water Act____Safe Drinking Water Act__
[1.14548](https://www.sigmaaldrich.com/US/en/product/mm/114548)Sulfate Cell Test5 - 250 mg/L SO4Yes
[1.14564](https://www.sigmaaldrich.com/US/en/product/mm/114564)Sulfate Cell Test100 - 1,000 mg/L SO4Yes
__Related Articles__
- [Comprehensive Quality Assurance and Process Control](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/analytical-chemistry/photometry-and-reflectometry/comprehensive-quality-assurance-and-process-control)
- [Determination of Bromate in Water and Drinking Water](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/analytical-chemistry/photometry-and-reflectometry/determination-of-bromate-in-water-and-drinking-water)
- [Efficient PAH Analysis (EU 15+1) using an Ascentis® Express PAH HPLC Column](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/analytical-chemistry/small-molecule-hplc/efficient-pah-analysis)
- [Ion Chromatography for Trace Cation Analysis](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/analytical-chemistry/small-molecule-hplc/ion-chromatography-and-cation-suppression-straightforward-cation-analysis-at-trace-levels)
- [PFAS Analysis Methods](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/analytical-chemistry/small-molecule-hplc/pfas-analysis-methods)
- [PFAS Column Selection for LC and GC](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/analytical-chemistry/small-molecule-hplc/pfas-column-selection-for-lc-and-gc)
- [Calculating Your Working Tolerance: A Guideline](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/environmental-testing-and-industrial-hygiene/drinking-water-testing/crm-calculating-working-tolerance)
- [Determination of 7 Nitrosamines in Drinking Water](https://www.sigmaaldrich.com/US/en/technical-documents/technical-article/environmental-testing-and-industrial-hygiene/drinking-water-testing/determination-of-7-nitrosamines-in-drinking-water)
- [View More](https://www.sigmaaldrich.com/US/en/search/facet-search?focus=sitecontent&term=facet-search)
__Related Product Categories__
- [Photometric analysis with Spectroquant® instruments and test kits](https://www.sigmaaldrich.com/US/en/products/analytical-chemistry/photometry-and-rapid-chemical-testing/photometry-instruments-kits-and-accessories)
- [Organic Building Blocks](https://www.sigmaaldrich.com/US/en/products/chemistry-and-biochemicals/building-blocks/organic-building-blocks)
- [Colorimetric and titrimetric test kits](https://www.sigmaaldrich.com/US/en/products/analytical-chemistry/photometry-and-rapid-chemical-testing/colorimetric-and-titrimetric-test-kits)
- [Photometry & rapid chemical testing](https://www.sigmaaldrich.com/US/en/products/analytical-chemistry/photometry-and-rapid-chemical-testing)
Top
__Sign In To Continue__
Sign In__Don't Have An Account?__Register
Support[Customer Support](https://www.sigmaaldrich.com/US/en/support/customer-support)[Contact Us](https://www.sigmaaldrich.com/US/en/collections/offices)[FAQ](https://maestro.my.site.com/knowledgeportal/s/)[Safety Data Sheets (SDS)](https://www.sigmaaldrich.com/US/en/documents-search?tab=sds)[Certificates (COA/COO)](https://www.sigmaaldrich.com/US/en/documents-search?tab=coa)[Quality & Regulatory](https://www.sigmaaldrich.com/US/en/life-science/quality-and-regulatory-management)[Calculators & Apps](https://www.sigmaaldrich.com/US/en/support/calculators-and-apps)[Webinars](https://www.sigmaaldrich.com/US/en/collections/webinars)
Orders[Order Lookup](https://www.sigmaaldrich.com/US/en/order-lookup)[Quick Order](https://www.sigmaaldrich.com/US/en/quick-order)[Custom Products](https://www.sigmaaldrich.com/US/en/services/custom-products)[eCommerce Solutions](https://www.sigmaaldrich.com/US/en/life-science/ecommerce/ecommerce-solutions)
Company[About Us](https://www.sigmaaldrich.com/US/en/life-science/about-us)[Responsibility](https://www.sigmaaldrich.com/US/en/life-science/ssbi)[Events](https://www.sigmaaldrich.com/US/en/collections/events)[Press Releases](https://www.sigmaaldrich.com/US/en/collections/press)[Programs](https://www.sigmaaldrich.com/US/en/life-science/partnership-programs)[Careers](https://careers.merckgroup.com/global/en)[Offices](https://www.sigmaaldrich.com/US/en/collections/offices)
Social Media
[](https://www.linkedin.com/company/merck-life-science)
[](https://x.com/Merck_lifesci)
[](https://www.facebook.com/Merck.lifescience)
[](https://www.instagram.com/mercklifescience/)
[](https://www.youtube.com/@MerckLifeScience/featured)
MilliporeSigma
__Research. Development. Production.__
We are a leading supplier to the global Life Science industry with solutions and services for research, biotechnology development and production, and pharmaceutical drug therapy development and production.
[](https://www.sigmaaldrich.com/US/en)
[Sigma-Aldrich® Solutions](https://www.sigmaaldrich.com/US/en/life-science/sigma-aldrich)[BioReliance® Solutions](https://www.sigmaaldrich.com/US/en/life-science/bioreliance)[Millipore® Solutions](https://www.sigmaaldrich.com/US/en/life-science/millipore)[SAFC® Solutions](https://www.sigmaaldrich.com/US/en/life-science/safc)[Milli-Q® Solutions](https://www.sigmaaldrich.com/US/en/life-science/milliq)[Supelco® Solutions](https://www.sigmaaldrich.com/US/en/life-science/supelco)
© 2026 Merck KGaA, Darmstadt, Germany and/or its affiliates. All Rights Reserved, including Text and Data Mining for AI training and similar technologies.
Reproduction of any materials from the site is strictly forbidden without permission.
[Site Use Terms](https://www.sigmaaldrich.com/US/en/life-science/legal/site-use-terms)|[Privacy Policy](https://www.sigmaaldrich.com/US/en/life-science/legal/privacy-statement)|[General Terms and Conditions of Sale](https://www.sigmaaldrich.com/US/en/life-science/legal/terms-and-conditions)|[Copyright Consent](https://www.sigmaaldrich.com/US/en/life-science/legal/copyright-consent) |[Site Map](https://www.sigmaaldrich.com/US/en/site-map)