Introducing Supel™ QuE Z-Sep Sorbents for QuEChERS
QuEChERs is a solid phase extraction (SPE) technique for sample preparation. Supelco's innovative Z-Sep and  Z-Sep+ sorbents demonstrated here extract more fat and pigment than traditional PSA/C18 phases while showing greater analyte recovery and greater reproducibility. Z-Sep sorbents are inexpensive, safe and easy to use.
For more on Supel™ QuE Z-Sep QuEChERS Tubes, click here.