Life Science
Bio Brief: Hybri-Max DMSO - D2650
Sigma's Hybri-Max™ DMSO is supplied in single-use, flame-sealed ampules, eliminating the need for time-consuming filtration and preventing microbial contamination. This is the researcher-preferred DMSO cryoprotectant for cell culture freezing medium.
  • Sterile-filtered and endotoxin-tested
  • Sealed ampules: 5 mL and 10 mL available
  • Suitable for hybridoma & cell freezing
  • Product Number D2650
Hybri-Max is a trademark of Sigma-Aldrich Co. LLC.