GenElute™ HP Plasmid Maxiprep procedure
The GenElute™ HP Plasmid Maxiprep kit offers a simple, rapid, and cost-effective method for isolating plasmid DNA from recombinant E. coli cutures. The recovered plasmid DNA is ready for immediate use in downstream applications, such as restriction digestion, ligation, sequencing, PCR, transformation, and transfection.
  • Culture Volume - 150 mL bacterial culture
  • Yield - Up to 1.2 mg yield of high-copy plasmid DNA
  • Speed - From harvested bacterial culture to pure plasmid DNA in 30 minutes or less
  • Flexible - Offers the flexibility of a vacuum or spin format, does not rely on gravity flow or ultracentrifugation
  • Economic - Lower cost per prep plus higher yields per prep equals double savings
Materials