GenElute™ Five-Minute Plasmid Miniprep procedure
The GenElute™ Five-Minute Plasmid Miniprep kit features an ultra streamlined protocol yielding high-quality plasmid DNA in about five minutes. The DNA is suitable for immediate use in capillary DNA sequencing, clone screening, restriction digestion and PCR.
  • Sophisticated – No pelleting cells or clearing lysates
  • Flexible – Binding column works with any standard laboratory vacuum manifold
  • Plentiful – Up to 5 µg of plasmid DNA or enough for 15 sequencing reactions
Materials