Choosing Thread Adapters for Dispensette® Dispensers
BrandTech Scientific, Inc. offers a wide range of thread adapters made of polypropylene, ETFE, or PTFE. Brand thread adapters fit all common thread finishes. For some type of bottles, such as the bottles with standard tapered joint, thread adapters may need to be stacked.
Materials