Choosing Thread Adapters for BRAND® Dispensette® S
This video provides an overview of how to make sure you pick the right thread adapters for your BRAND® Dispensette® S bottle-top dispenser. The BRAND® Dispensette® S bottle-top dispensers come with different models to dispense virtually laboratory liquids such as concentrated acids, difficult organic solvents and high purity reagents. BRAND® Dispensette® S bottle-top dispensers contains an improved recirculation valve, a hinged safety cap, smoother and lower force operation, an improved safety discharge system, and a washer-free sealing system.
Materials