Transferpette® S Pipettes from BrandTech® Scientific
Transferpette® S Pipettes a manufactured in Germany. Both single-channel and multi-channel models are available in volumes ranging from 0.1 µL to 10 µL and 0.5 µL to 300 µL, respectively. The user friendly features include short pipetting stroke and large separate tip ejector. Transferpette® S’ nose cone accepts other manufacturers’ tips. The color-coded pipettes also have name plates. The maintenance and calibration can be easily done by the end-user.
Materials