Cellometer Auto 2000 from Nexcelcom
The Cellometer Auto 2000 utilizes bright-field and dual-fluorescence imaging to quickly identify and count individual cells with a high level of accuracy. Cell count, concentration, diameter, and percent viability are automatically calculated and reported. With the click of a button, researchers can select a new assay with saved counting parameters, making the instrument ideal for labs testing a variety of cell types.