Whatman™ Glass Mini-UniPrep™ G2 Syringeless Filters
The Mini-UniPrep G2 syringeless filters from Whatman provide a fast and easy method of removing particulates from HPLC and uHPLC samples. These innovative units can lower sample preparation time to one third of that required with other methods, minimize the presence of leachables when compared to standard, plastic filters and will allow you to cut several consumable pieces out of your inventory. These preassembled, glass syringeless filtration devices must be used with a compressor unit.
Browse all filtration products.