Wrap-It Tie Installation
The self-locking, one-piece nylon construction of the Wrap-it tie provides an excellent way to secure rubber septa to bottles and flask joints. By using the Wrap-it tie and the installing tool, easy attachment of tubing to hose barbs or glassware, is accomplished. Wrap-it ties are easily removed with the All-in-One tool.