Elix® EDI Module for Water Purification Systems
Many of our Type 2 pure water systems, including the new Milli-Q® IX pure water system, incorporate the unique Elix® electrodeionization (EDI) module. This environmentally-friendly technology is based on electrochemical regeneration, which replaces the chemical regeneration of conventional ion-exchange systems and also removes the need for chemical or resin disposal. With over 25 years of proven success, Elix® pure water is suited to every user’s needs in today’s demanding laboratory environment. Learn More