Attention:
We are currently upgrading the certain systems that support this website.  You may experience some functionality loss, though we expect the site to be fully functional by 9:00PM CT (US), February 24th.
We apologize for any inconvenience and appreciate your patience during this improvement process.Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Live Cell Imaging of Autophagy using LentiBrite™ Fluorescent Biosensors
HT-1080 cells were plated in a chamber slide and transduced with LentiBrite™ GFP-LC3 lentiviral particles at an MOI of 20 for 24 hours. After media replacement and 48 hours further incubation, cells were incubated for 4 hours in EBSS containing a lysosome inhibitor, to induce autophagy and inhibit lysosomal degradation of autophagosomes. LentiBrite™ Lentiviral Biosensors are a new suite of pre-packaged lentiviral particles encoding important and foundational proteins of autophagy, apoptosis, and cell structure for visualization under different cell/disease states in live cell and in vitro analysis.
Materials