Materials Science
ALD Cylinders for Pyrophoric Precursors
Our Material Science portfolio offers a variety of precursor materials for use in Atomic Layer Deposition, or ALD. ALD uses consecutive, self-limiting reactions to deposit thin films of controlled thickness, uniformity, and chemical composition.