Sign In to View Organizational & Contract Pricing
About This Item
Empirical Formula (Hill Notation):
C20H22O6
CAS Number:
Molecular Weight:
358.39
Beilstein/REAXYS Number:
7826317
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:
NACRES:
NA.21
Skip To
Quality Level
assay
≥85% (HPLC)
storage temp.
2-8°C
SMILES string
COc1cc(C[C@H]2COC(=O)[C@@H]2Cc3ccc(O)c(OC)c3)ccc1O
InChI
1S/C20H22O6/c1-24-18-9-12(3-5-16(18)21)7-14-11-26-20(23)15(14)8-13-4-6-17(22)19(10-13)25-2/h3-6,9-10,14-15,21-22H,7-8,11H2,1-2H3/t14-,15+/m0/s1
Inchi Key
MATGKVZWFZHCLI-LSDHHAIUSA-N
Looking for similar products? Visit Product Comparison Guide
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
L.P. Meagher, G.R. Beecher
J. Food Compos. Anal., 13, 935-935 (2000)
Tanja Kraushofer et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 777(1-2), 61-66 (2002-09-25)
A HPLC method coupled with coulometric electrode array detection for the determination of matairesinol in flax seed is described. The defatted sample was spiked with bisphenol A (internal standard), refluxed for 75 min in a mixture of ethanol-bidistilled water-12 M
Boram Lee et al.
Biochemical and biophysical research communications, 421(1), 76-80 (2012-04-10)
Mitochondrial reactive oxygen species (mROS) are involved in cancer initiation and progression and function as signaling molecules in many aspects of hypoxia and growth factor-mediated signaling. Here we report that matairesinol, a natural small molecule identified from the cell-based screening
Saya Kawahara et al.
Bioscience, biotechnology, and biochemistry, 74(9), 1878-1883 (2010-09-14)
The IgE-suppressive activity of (-)-matairesinol is demonstrated, and the structure-activity relationship of (-)-matairesinol clarified. 3',4-Dihydroxy-3,4'-dimethoxylignano-9,9'-lactone showed higher IgE-suppressive activity than (-)-matairesinol without any cytotoxic activity. Some derivatives bearing a longer and more bulky alkoxy group at the 3 or 4
Johanna W Lampe et al.
Journal of AOAC International, 89(4), 1174-1181 (2006-08-19)
Phytoestrogens occur naturally in plants and are structurally similar to mammalian estrogens. The lignans are a class of phytoestrogen and can be metabolized to the biologically active enterolignans, enterodiol, and enterolactone by a consortium of intestinal bacteria. Secoisolariciresinol diglucoside (SDG)
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service