Skip to Content

BDPH9BCC1C19

Bide Pharmatech

2-Chloro-6-methyl-4-(trifluoromethyl)nicotinonitrile

Synonym(s):

2-chloro-6-methyl-4-(trifluoromethyl)nicotinonitrile

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C8H4N2Cl1F3
CAS Number:
Molecular Weight:
220.58
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C8H4ClF3N2/c1-4-2-6(8(10,11)12)5(3-13)7(9)14-4/h2H,1H3

Inchi Key

LNJOJFNFYGSCCR-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Liquid



Still not finding the right product?


Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library