Skip to Content
Merck

BDPH9BCC3BD5

Bide Pharmatech

2-Chloro-4-(tributylstannyl)pyrimidine

Synonym(s):

2-chloro-4-(tributylstannyl)pyrimidine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C16H29N2Cl1Sn1
Molecular Weight:
403.58
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C4H2ClN2.3C4H9.Sn/c5-4-6-2-1-3-7-4;3*1-3-4-2;/h1-2H;3*1,3-4H2,2H3;

Inchi Key

RYGIFHOTRLEMGG-UHFFFAOYSA-N





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library