Skip to Content
Merck

BDPH9BCC88DE

Bide Pharmatech

(3-Chlorophenyl)carbonohydrazonoyl dicyanide

Synonym(s):

2-[(3-chlorophenyl)hydrazono]malononitrile

Select a Size

Change View

About This Item

Linear Formula:
C9H5N4Cl1
CAS Number:
Molecular Weight:
204.62
MDL number:
Sign in to see your pricing
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C9H5ClN4/c10-7-2-1-3-8(4-7)13-14-9(5-11)6-12/h1-4,13H

Inchi Key

UGTJLJZQQFGTJD-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

99%

Storage temp.

Keep in dark place, inert atmosphere, room temperature

Country of Origin

CN

Physical Form

Form: powder



Still not finding the right product?



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library