Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
Linear Formula:
C12H8O1N1Cl1Br1F1
CAS Number:
Molecular Weight:
316.56
MDL number:
Pricing and availability is not currently available. Contact Technical Service
Inchi Code
1S/C12H8BrClFNO/c13-11-2-1-3-12(16-11)17-7-8-4-5-9(14)6-10(8)15/h1-6H,7H2
Inchi Key
JCOBZUDDQNLDTA-UHFFFAOYSA-N
Shipping Temperature
Normal
Purity
95%
Storage temp.
Inert atmosphere, 2-8C
Country of Origin
CN
Physical Form
White solid
Still not finding the right product?
Explore all of our products under 2-Bromo-6-((4-chloro-2-fluorobenzyl)oxy)pyridine
hstatements
signalword
Warning
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
