Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
Pricing and availability is not currently available. Contact Technical Service
Inchi Code
1S/C6H5ClN4/c7-4-2-1-3-5-9-6(8)10-11(4)5/h1-3H,(H2,8,10)
Inchi Key
RCCCLZDQZNFDGK-UHFFFAOYSA-N
Shipping Temperature
Normal
Purity
98%
Storage temp.
Keep in dark place, inert atmosphere, 2-8C
Country of Origin
CN
Physical Form
Solid
Still not finding the right product?
Explore all of our products under 5-Chloro-[1,2,4]triazolo[1,5-a]pyridin-2-amine
hstatements
Precautionary Statements
Signal Word
Warning
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
