Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
Pricing and availability is not currently available. Contact Technical Service
Inchi Code
1S/C8H7ClO5S/c1-14-6-3-2-5(8(10)11)4-7(6)15(9,12)13/h2-4H,1H3,(H,10,11)
Inchi Key
OWBBPHXIAFVUMO-UHFFFAOYSA-N
Shipping Temperature
Normal
Purity
95%
Storage temp.
Inert atmosphere, 2-8C
Country of Origin
CN
Physical Form
White to off-white powder or crystals
Still not finding the right product?
Explore all of our products under 3-(Chlorosulfonyl)-4-methoxybenzoic acid
hstatements
Precautionary Statements
Signal Word
Danger
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
