Skip to Content

BDPH9BCD2FB7

Bide Pharmatech

4-Bromo-1-chloro-2-(4-ethoxybenzyl)benzene

Synonym(s):

4-bromo-1-chloro-2-(4-ethoxybenzyl)benzene

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C15H14O1Cl1Br1
CAS Number:
Molecular Weight:
325.63
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C15H14BrClO/c1-2-18-14-6-3-11(4-7-14)9-12-10-13(16)5-8-15(12)17/h3-8,10H,2,9H2,1H3

Inchi Key

ZUNCHZBITMUSRD-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

White to off-white powder or crystals



Still not finding the right product?


hstatements

Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library