Skip to Content
Merck

BDPH9BCD4DEA

Bide Pharmatech

6-Iodochroman-2-carboxylic acid

Synonym(s):

6-iodo-2-chromanecarboxylic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C10H9O3I1
CAS Number:
Molecular Weight:
304.08
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C10H9IO3/c11-7-2-4-8-6(5-7)1-3-9(14-8)10(12)13/h2,4-5,9H,1,3H2,(H,12,13)

Inchi Key

KUUOJGIUGRFQCR-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Keep in dark place, sealed in dry, 2-8C

Country of Origin

CN

Physical Form

Powder or crystals



Still not finding the right product?

Explore all of our products under 6-Iodochroman-2-carboxylic acid




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library