Skip to Content
Merck

BDPH9BCD4F36

Bide Pharmatech

8-Chloroquinolin-6-ol

Synonym(s):

8-chloro-6-quinolinol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C9H6O1N1Cl1
CAS Number:
Molecular Weight:
179.61
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C9H6ClNO/c10-8-5-7(12)4-6-2-1-3-11-9(6)8/h1-5,12H

Inchi Key

NSFZSTFINAEFAB-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Inert atmosphere, room temperature

Country of Origin

CN

Physical Form

Off-white to gray to yellow powder or crystals



Still not finding the right product?

Explore all of our products under 8-Chloroquinolin-6-ol


hstatements

Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library