Skip to Content
Merck

BDPH9BCD53BF

Bide Pharmatech

1-(3-Bromo-5-methylphenyl)ethan-1-amine hydrochloride

Synonym(s):

1-(3-bromo-5-methylphenyl)ethan-1-amine hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C9H13N1Cl1Br1
CAS Number:
Molecular Weight:
250.57
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C9H12BrN.ClH/c1-6-3-8(7(2)11)5-9(10)4-6;/h3-5,7H,11H2,1-2H3;1H

Inchi Key

NNYNPWGFQJGLIN-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Inert atmosphere, room temperature

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library