Skip to Content

BDPH9BCD66DA

Bide Pharmatech

3-Bromo-2-(methylthio)-5-(trifluoromethyl)pyridine

Synonym(s):

3-bromo-5-(trifluoromethyl)-2-pyridinyl methyl sulfide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C7H5N1Br1S1F3
CAS Number:
Molecular Weight:
272.09
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C7H5BrF3NS/c1-13-6-5(8)2-4(3-12-6)7(9,10)11/h2-3H,1H3

Inchi Key

PGEREEIUJMZQTJ-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library