Skip to Content
Merck

BDPH9BCD76C3

Bide Pharmatech

2-(4-Iodophenyl)ethanol

Synonym(s):

2-(4-iodophenyl)ethanol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C8H9O1I1
CAS Number:
Molecular Weight:
248.06
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C8H9IO/c9-8-3-1-7(2-4-8)5-6-10/h1-4,10H,5-6H2

Inchi Key

GYUSTTSSRXDFKG-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Keep in dark place, sealed in dry, 2-8C

Country of Origin

CN

Physical Form

Colorless or white to yellow solid or semi-solid or liquid or lump



Still not finding the right product?

Explore all of our products under 2-(4-Iodophenyl)ethanol


hstatements

Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library