Skip to Content
Merck

BDPH9BCD948B

Bide Pharmatech

6-(Diethylamino)nicotinaldehyde

Synonym(s):

6-(diethylamino)nicotinaldehyde

Select a Size

Change View

About This Item

Linear Formula:
C10H14O1N2
CAS Number:
Molecular Weight:
178.23
MDL number:
Sign in to see your pricing
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C10H14N2O/c1-3-12(4-2)10-6-5-9(8-13)7-11-10/h5-8H,3-4H2,1-2H3

Inchi Key

PCISUIMHLSRKGR-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Solid or semi-solid or lump or liquid



Still not finding the right product?

Explore all of our products under 6-(Diethylamino)nicotinaldehyde


hstatements

Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library