Skip to Content
Merck

BDPH9BCD9AFE

Bide Pharmatech

Triisopropoxy(vinyl)silane

Synonym(s):

triisopropoxy(vinyl)silane

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C11H24O3Si1
Molecular Weight:
232.4
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C11H24O3Si/c1-8-15(12-9(2)3,13-10(4)5)14-11(6)7/h8-11H,1H2,2-7H3

Inchi Key

MABAWBWRUSBLKQ-UHFFFAOYSA-N





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library