Skip to Content
Merck

BDPH9BCDADC4

Bide Pharmatech

3-Bromo-1-methyl-1H-pyrazole-4-carboxylic acid

Synonym(s):

3-bromo-1-methyl-1H-pyrazole-4-carboxylic acid

Select a Size

Change View

About This Item

Linear Formula:
C5H5O2N2Br1
CAS Number:
Molecular Weight:
205.01
MDL number:
Sign in to see your pricing
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C5H5BrN2O2/c1-8-2-3(5(9)10)4(6)7-8/h2H,1H3,(H,9,10)

Inchi Key

LJOMSWUFAYFYEA-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Country of Origin

CN

Physical Form

White to Yellow Powder or crystals

Storage temp.

Inert atmosphere, 2-8C



Still not finding the right product?


Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library