Skip to Content
Merck

BDPH9BCDB127

Bide Pharmatech

Methyl 5-methyl-1H-pyrrole-2-carboxylate

Synonym(s):

methyl 5-methyl-1H-pyrrole-2-carboxylate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C7H9O2N1
CAS Number:
Molecular Weight:
139.15
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C7H9NO2/c1-5-3-4-6(8-5)7(9)10-2/h3-4,8H,1-2H3

Inchi Key

CQYALQAMQXUBRO-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

96%

Storage temp.

Inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

White to Yellow or Pale-brown to Brown Solid



Still not finding the right product?


Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library