Skip to Content
Merck

BDPH9BCDC239

Bide Pharmatech

Ethyl 5-chloro-3-(methylthio)-1,2,4-triazine-6-carboxylate

Synonym(s):

ethyl 5-chloro-3-(methylsulfanyl)-1,2,4-triazine-6-carboxylate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C7H8O2N3Cl1S1
CAS Number:
Molecular Weight:
233.68
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C7H8ClN3O2S/c1-3-13-6(12)4-5(8)9-7(14-2)11-10-4/h3H2,1-2H3

Inchi Key

WXJHHRMJGBPYEG-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Country of Origin

CN

Physical Form

Solid

Storage temp.

Inert atmosphere, 2-8°C



Still not finding the right product?


Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library