Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
Pricing and availability is not currently available. Contact Technical Service
Inchi Code
1S/C10H10N2O/c1-13-10-7-3-2-6-12-9(7)5-4-8(10)11/h2-6H,11H2,1H3
Inchi Key
RJUWMZNBSHEASH-UHFFFAOYSA-N
Shipping Temperature
Normal
Purity
97%
Storage temp.
Keep in dark place, inert atmosphere, room temperature
Country of Origin
CN
Physical Form
Solid or Semi-solid or liquid or lump
Still not finding the right product?
Explore all of our products under 5-Methoxyquinolin-6-amine
hstatements
Precautionary Statements
Signal Word
Warning
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
