Skip to Content

BDPH9BCDC80B

Bide Pharmatech

Ethyl 5-methyl-1,3,4-thiadiazole-2-carboxylate

Synonym(s):

ethyl 5-methyl-1,3,4-thiadiazole-2-carboxylate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C6H8O2N2S1
CAS Number:
Molecular Weight:
172.21
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C6H8N2O2S/c1-3-10-6(9)5-8-7-4(2)11-5/h3H2,1-2H3

Inchi Key

VMVKZLLIEGQHEE-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Sealed in dry, 2-8C

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?


hstatements

Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library