Skip to Content

BDPH9BCDCA79

Bide Pharmatech

6-Ethylpyridin-3-amine

Synonym(s):

6-ethyl-3-pyridinamine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C7H10N2
CAS Number:
Molecular Weight:
122.17
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C7H10N2/c1-2-7-4-3-6(8)5-9-7/h3-5H,2,8H2,1H3

Inchi Key

ZKPKEODUAJQPCV-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Keep in dark place, inert atmosphere, room temperature

Country of Origin

CN

Physical Form

Liquid or Solid or Semi-solid



Still not finding the right product?

Explore all of our products under 6-Ethylpyridin-3-amine


hstatements

Precautionary Statements

Signal Word

Danger

Pictograms

Corrosion


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library